Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 56381485-56381612 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AURKA
Entrez Gene ID
6790
Gene Name
aurora kinase A
Gene Aliases
AIK, ARK1, AURA, BTAK, PPP1R47, STK15, STK6, STK7
UniGene
Hs.250822
Gene Chromosome Location
Chr.20: 56369389-56392337 on Build GRCh38
Transcripts
NM_001323303.1 NM_001323304.1 NM_001323305.1 NM_003600.3 NM_198433.2 NM_198434.2 NM_198435.2 NM_198436.2 NM_198437.2 XM_017028034.1 XM_017028035.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1751509 c.624A>G p.L208L Chr.20: 56381514-56381514 on Build GRCh38
6913385 c.652G>T p.V218F Chr.20: 56381486-56381486 on Build GRCh38
4099634 c.617T>C p.V206A Chr.20: 56381521-56381521 on Build GRCh38
3548023 c.594T>G p.G198G Chr.20: 56381544-56381544 on Build GRCh38
6845572 c.571C>T p.P191S Chr.20: 56381567-56381567 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750995293 Chr.20: 56381576-56381576 on Build GRCh38
rs534866768 Chr.20: 56381517-56381517 on Build GRCh38
rs45483697 Chr.20: 56381546-56381546 on Build GRCh38
rs765786653 Chr.20: 56381575-56381575 on Build GRCh38
rs762388330 Chr.20: 56381545-56381545 on Build GRCh38
rs972022489 Chr.20: 56381548-56381548 on Build GRCh38
rs776320107 Chr.20: 56381537-56381537 on Build GRCh38

View Full Product Details