Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 152323074-152323207 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXW7
Entrez Gene ID
55294
Gene Name
F-box and WD repeat domain containing 7
Gene Aliases
AGO, CDC4, FBW6, FBW7, FBX30, FBXO30, FBXW6, SEL-10, SEL10, hAgo, hCdc4
UniGene
Hs.561245
Gene Chromosome Location
Chr.4: 152321258-152536095 on Build GRCh38
Transcripts
NM_001013415.1 NM_018315.4 NM_033632.3 XM_011532083.1 XM_011532084.1 XM_011532085.1 XM_011532086.1 XM_011532087.1 XM_011532088.1 XM_017008362.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6939426 c.1532_1535delGTTT p.C511fs*8 Chr.4: 152323116-152323119 on Build GRCh38
5344633 c.1931G>A p.G644E Chr.4: 152323074-152323074 on Build GRCh38
5432179 c.1616-2A>G p.? Chr.4: 152323151-152323151 on Build GRCh38
6925218 c.1191T>A p.F397L Chr.4: 152323097-152323097 on Build GRCh38
6662534 c.1556T>C p.V519A Chr.4: 152323095-152323095 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766010404 Chr.4: 152323139-152323139 on Build GRCh38
rs376275227 Chr.4: 152323153-152323153 on Build GRCh38
rs375535587 Chr.4: 152323161-152323161 on Build GRCh38
rs780673225 Chr.4: 152323167-152323167 on Build GRCh38
rs954236185 Chr.4: 152323155-152323155 on Build GRCh38
rs758163453 Chr.4: 152323107-152323107 on Build GRCh38
rs762244922 Chr.4: 152323158-152323158 on Build GRCh38

View Full Product Details