Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 7870777-7870950 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTRR
Entrez Gene ID
4552
Gene Name
5-methyltetrahydrofolate-homocysteine methyltransferase reductase
Gene Aliases
MSR, cblE
UniGene
Hs.481551
Gene Chromosome Location
Chr.5: 7869104-7901124 on Build GRCh38
Transcripts
NM_002454.2 NM_024010.2 NR_134480.1 NR_134481.1 NR_134482.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5810395 c.104A>G p.Y35C Chr.5: 7870817-7870817 on Build GRCh38
4848818 c.76G>A p.E26K Chr.5: 7870789-7870789 on Build GRCh38
6581170 c.94C>G p.L32V Chr.5: 7870807-7870807 on Build GRCh38
739140 c.84G>T p.M28I Chr.5: 7870797-7870797 on Build GRCh38
6716965 c.135C>T p.I45I Chr.5: 7870848-7870848 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs560414393 Chr.5: 7870816-7870816 on Build GRCh38
rs779667897 Chr.5: 7870807-7870812 on Build GRCh38
rs6413426 Chr.5: 7870848-7870848 on Build GRCh38
rs752509731 Chr.5: 7870833-7870833 on Build GRCh38
rs756074729 Chr.5: 7870835-7870835 on Build GRCh38
rs771299380 Chr.5: 7870894-7870894 on Build GRCh38
rs767503539 Chr.5: 7870824-7870824 on Build GRCh38
rs114748706 Chr.5: 7870821-7870821 on Build GRCh38
rs748657512 Chr.5: 7870867-7870867 on Build GRCh38
rs781726691 Chr.5: 7870861-7870861 on Build GRCh38
rs769772796 Chr.5: 7870852-7870852 on Build GRCh38
rs775343389 Chr.5: 7870919-7870919 on Build GRCh38
rs114427781 Chr.5: 7870815-7870815 on Build GRCh38
rs749337272 Chr.5: 7870883-7870883 on Build GRCh38
rs1801394 Chr.5: 7870860-7870860 on Build GRCh38
rs751400453 Chr.5: 7870820-7870820 on Build GRCh38
rs763313419 Chr.5: 7870807-7870807 on Build GRCh38
rs954239819 Chr.5: 7870850-7870850 on Build GRCh38
rs777323334 Chr.5: 7870837-7870837 on Build GRCh38
rs747426239 Chr.5: 7870857-7870857 on Build GRCh38
rs759379396 Chr.5: 7870917-7870917 on Build GRCh38
rs141118068 Chr.5: 7870895-7870895 on Build GRCh38
rs937595017 Chr.5: 7870921-7870921 on Build GRCh38
rs753325140 Chr.5: 7870846-7870846 on Build GRCh38
rs1045121482 Chr.5: 7870878-7870878 on Build GRCh38
rs773463459 Chr.5: 7870877-7870877 on Build GRCh38
rs767281477 Chr.5: 7870918-7870918 on Build GRCh38
rs748064430 Chr.5: 7870843-7870843 on Build GRCh38
rs114847469 Chr.5: 7870849-7870849 on Build GRCh38

View Full Product Details