Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 133098531-133098697 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RALGDS
Entrez Gene ID
5900
Gene Name
ral guanine nucleotide dissociation stimulator
Gene Aliases
RGDS, RGF, RalGEF
UniGene
Hs.106185
Gene Chromosome Location
Chr.9: 133097720-133149220 on Build GRCh38
Transcripts
NM_001042368.2 NM_001271774.1 NM_001271775.1 NM_001271776.1 NM_006266.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4418889 c.2853C>G p.L951L Chr.9: 133098692-133098692 on Build GRCh38
7182635 c.2952C>T p.I984I Chr.9: 133098593-133098593 on Build GRCh38
4371733 c.2645G>A p.R882Q Chr.9: 133098684-133098684 on Build GRCh38
7182632 c.2739C>T p.I913I Chr.9: 133098593-133098593 on Build GRCh38
4720170 c.2631A>G p.G877G Chr.9: 133098614-133098614 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs568299176 Chr.9: 133098622-133098622 on Build GRCh38
rs764693552 Chr.9: 133098600-133098600 on Build GRCh38
rs370690294 Chr.9: 133098645-133098645 on Build GRCh38
rs370951865 Chr.9: 133098606-133098606 on Build GRCh38
rs746054003 Chr.9: 133098610-133098610 on Build GRCh38
rs773295357 Chr.9: 133098558-133098558 on Build GRCh38
rs1026856324 Chr.9: 133098631-133098631 on Build GRCh38
rs376030443 Chr.9: 133098642-133098642 on Build GRCh38
rs776277724 Chr.9: 133098572-133098572 on Build GRCh38
rs531302066 Chr.9: 133098583-133098583 on Build GRCh38
rs781302534 Chr.9: 133098609-133098609 on Build GRCh38
rs760651358 Chr.9: 133098573-133098587 on Build GRCh38
rs145020652 Chr.9: 133098653-133098653 on Build GRCh38
rs143209866 Chr.9: 133098667-133098667 on Build GRCh38
rs149083027 Chr.9: 133098666-133098666 on Build GRCh38
rs760169037 Chr.9: 133098558-133098558 on Build GRCh38
rs773078165 Chr.9: 133098654-133098654 on Build GRCh38
rs759013918 Chr.9: 133098597-133098597 on Build GRCh38
rs375124486 Chr.9: 133098661-133098661 on Build GRCh38
rs370328313 Chr.9: 133098563-133098563 on Build GRCh38
rs766914011 Chr.9: 133098609-133098610 on Build GRCh38
rs996566211 Chr.9: 133098628-133098628 on Build GRCh38
rs759279035 Chr.9: 133098677-133098677 on Build GRCh38
rs190743751 Chr.9: 133098602-133098602 on Build GRCh38
rs776167423 Chr.9: 133098673-133098673 on Build GRCh38
rs374088319 Chr.9: 133098582-133098582 on Build GRCh38
rs747935608 Chr.9: 133098652-133098652 on Build GRCh38
rs957917376 Chr.9: 133098559-133098559 on Build GRCh38
rs753085695 Chr.9: 133098596-133098596 on Build GRCh38

View Full Product Details