Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 41932122-41932274 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KAT6A
Entrez Gene ID
7994
Gene Name
lysine acetyltransferase 6A
Gene Aliases
MOZ, MRD32, MYST-3, MYST3, RUNXBP2, ZC2HC6A, ZNF220
UniGene
Hs.491577
Gene Chromosome Location
Chr.8: 41929479-42052026 on Build GRCh38
Transcripts
NM_006766.4 XM_011544656.2 XM_011544657.2 XM_011544658.2 XM_011544659.2 XM_017013863.1 XM_017013864.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4843404 c.5972G>A p.G1991D Chr.8: 41932248-41932248 on Build GRCh38
283084 c.5965G>A p.A1989T Chr.8: 41932255-41932255 on Build GRCh38
339908 c.5954G>A p.S1985N Chr.8: 41932266-41932266 on Build GRCh38
2961641 c.5973C>T p.G1991G Chr.8: 41932247-41932247 on Build GRCh38
2961642 c.5964C>T p.N1988N Chr.8: 41932256-41932256 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771900139 Chr.8: 41932237-41932237 on Build GRCh38
rs778809488 Chr.8: 41932227-41932227 on Build GRCh38
rs1050612463 Chr.8: 41932246-41932246 on Build GRCh38
rs376166628 Chr.8: 41932188-41932188 on Build GRCh38
rs753662262 Chr.8: 41932225-41932225 on Build GRCh38
rs746592247 Chr.8: 41932248-41932248 on Build GRCh38
rs760077026 Chr.8: 41932184-41932184 on Build GRCh38
rs547173239 Chr.8: 41932155-41932155 on Build GRCh38
rs751719703 Chr.8: 41932195-41932195 on Build GRCh38
rs777771800 Chr.8: 41932216-41932216 on Build GRCh38
rs373063742 Chr.8: 41932199-41932199 on Build GRCh38
rs749178614 Chr.8: 41932175-41932180 on Build GRCh38
rs777976735 Chr.8: 41932178-41932179 on Build GRCh38
rs117270099 Chr.8: 41932226-41932226 on Build GRCh38
rs750678051 Chr.8: 41932191-41932191 on Build GRCh38
rs768772608 Chr.8: 41932174-41932175 on Build GRCh38
rs745467008 Chr.8: 41932162-41932162 on Build GRCh38
rs971987999 Chr.8: 41932173-41932176 on Build GRCh38
rs745627517 Chr.8: 41932229-41932229 on Build GRCh38
rs771738778 Chr.8: 41932169-41932169 on Build GRCh38
rs779835230 Chr.8: 41932247-41932247 on Build GRCh38
rs764156028 Chr.8: 41932146-41932146 on Build GRCh38
rs914972486 Chr.8: 41932201-41932201 on Build GRCh38
rs778719613 Chr.8: 41932160-41932160 on Build GRCh38
rs775180392 Chr.8: 41932176-41932176 on Build GRCh38
rs753328385 Chr.8: 41932186-41932186 on Build GRCh38
rs765539082 Chr.8: 41932189-41932189 on Build GRCh38
rs974454875 Chr.8: 41932175-41932175 on Build GRCh38
rs749175939 Chr.8: 41932156-41932156 on Build GRCh38
rs143452263 Chr.8: 41932253-41932253 on Build GRCh38
rs766570372 Chr.8: 41932194-41932194 on Build GRCh38

View Full Product Details