Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 128151695-128151954 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LCN2
Entrez Gene ID
3934
Gene Name
lipocalin 2
Gene Aliases
24p3, MSFI, NGAL, p25
UniGene
Hs.204238
Gene Chromosome Location
Chr.9: 128149430-128153455 on Build GRCh38
Transcripts
NM_005564.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6295263 c.362-8G>T p.? Chr.9: 128151898-128151898 on Build GRCh38
5646100 c.376A>G p.T126A Chr.9: 128151920-128151920 on Build GRCh38
3904360 c.345G>A p.T115T Chr.9: 128151701-128151701 on Build GRCh38
7122515 c.387C>T p.L129L Chr.9: 128151931-128151931 on Build GRCh38
3326463 c.339G>A p.T113T Chr.9: 128151701-128151701 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201401236 Chr.9: 128151897-128151897 on Build GRCh38
rs921527809 Chr.9: 128151833-128151833 on Build GRCh38
rs772645506 Chr.9: 128151856-128151856 on Build GRCh38
rs548592990 Chr.9: 128151787-128151787 on Build GRCh38
rs972576279 Chr.9: 128151777-128151777 on Build GRCh38
rs934133059 Chr.9: 128151838-128151838 on Build GRCh38
rs773532605 Chr.9: 128151910-128151910 on Build GRCh38
rs537466053 Chr.9: 128151825-128151825 on Build GRCh38
rs910496781 Chr.9: 128151827-128151827 on Build GRCh38
rs756089088 Chr.9: 128151898-128151898 on Build GRCh38
rs986948574 Chr.9: 128151851-128151851 on Build GRCh38
rs116745581 Chr.9: 128151903-128151903 on Build GRCh38
rs762395225 Chr.9: 128151922-128151922 on Build GRCh38
rs139329518 Chr.9: 128151719-128151719 on Build GRCh38
rs756180761 Chr.9: 128151935-128151935 on Build GRCh38
rs375490659 Chr.9: 128151876-128151876 on Build GRCh38
rs749315676 Chr.9: 128151901-128151901 on Build GRCh38
rs11794980 Chr.9: 128151891-128151891 on Build GRCh38
rs566632836 Chr.9: 128151756-128151756 on Build GRCh38
rs757282474 Chr.9: 128151724-128151724 on Build GRCh38
rs762299912 Chr.9: 128151923-128151923 on Build GRCh38
rs908603171 Chr.9: 128151857-128151857 on Build GRCh38
rs1021326745 Chr.9: 128151775-128151775 on Build GRCh38
rs879083145 Chr.9: 128151905-128151906 on Build GRCh38
rs780332123 Chr.9: 128151900-128151900 on Build GRCh38
rs780457081 Chr.9: 128151738-128151738 on Build GRCh38
rs749567379 Chr.9: 128151743-128151743 on Build GRCh38
rs200328232 Chr.9: 128151895-128151895 on Build GRCh38
rs763824629 Chr.9: 128151925-128151925 on Build GRCh38
rs372560040 Chr.9: 128151765-128151765 on Build GRCh38
rs141218430 Chr.9: 128151932-128151932 on Build GRCh38
rs752177802 Chr.9: 128151882-128151882 on Build GRCh38
rs376581902 Chr.9: 128151846-128151846 on Build GRCh38
rs975598738 Chr.9: 128151818-128151818 on Build GRCh38
rs570958879 Chr.9: 128151861-128151861 on Build GRCh38
rs762016178 Chr.9: 128151763-128151763 on Build GRCh38
rs1002422983 Chr.9: 128151929-128151929 on Build GRCh38
rs762720292 Chr.9: 128151890-128151890 on Build GRCh38
rs919244629 Chr.9: 128151790-128151790 on Build GRCh38
rs2232626 Chr.9: 128151725-128151725 on Build GRCh38
rs2232627 Chr.9: 128151911-128151911 on Build GRCh38
rs183777464 Chr.9: 128151736-128151736 on Build GRCh38
rs200300117 Chr.9: 128151757-128151757 on Build GRCh38
rs745961653 Chr.9: 128151733-128151733 on Build GRCh38
rs553580716 Chr.9: 128151902-128151902 on Build GRCh38
rs557457023 Chr.9: 128151826-128151826 on Build GRCh38
rs761702759 Chr.9: 128151928-128151928 on Build GRCh38
rs568419305 Chr.9: 128151794-128151794 on Build GRCh38
rs772384732 Chr.9: 128151908-128151908 on Build GRCh38
rs756947984 Chr.9: 128151892-128151892 on Build GRCh38
rs79993583 Chr.9: 128151921-128151921 on Build GRCh38
rs773829949 Chr.9: 128151926-128151926 on Build GRCh38
rs943272085 Chr.9: 128151830-128151830 on Build GRCh38
rs759075809 Chr.9: 128151872-128151872 on Build GRCh38

View Full Product Details