Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 74636461-74636708 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RFWD3
Entrez Gene ID
55159
Gene Name
ring finger and WD repeat domain 3
Gene Aliases
RNF201
UniGene
Hs.567525
Gene Chromosome Location
Chr.16: 74621396-74666892 on Build GRCh38
Transcripts
NM_018124.3 XM_005256021.4 XM_005256022.4 XM_006721228.3 XM_011523191.2 XM_017023391.1 XM_017023392.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3888953 c.1280C>T p.S427F Chr.16: 74636492-74636492 on Build GRCh38
5981030 c.1297A>G p.K433E Chr.16: 74636475-74636475 on Build GRCh38
704146 c.1195-2A>C p.? Chr.16: 74636579-74636579 on Build GRCh38
5730916 c.1248C>T p.G416G Chr.16: 74636524-74636524 on Build GRCh38
4062773 c.1293G>A p.Q431Q Chr.16: 74636479-74636479 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770295813 Chr.16: 74636497-74636497 on Build GRCh38
rs1032864186 Chr.16: 74636661-74636661 on Build GRCh38
rs113755607 Chr.16: 74636576-74636576 on Build GRCh38
rs747018311 Chr.16: 74636561-74636561 on Build GRCh38
rs746186531 Chr.16: 74636486-74636486 on Build GRCh38
rs1007558452 Chr.16: 74636655-74636655 on Build GRCh38
rs746377038 Chr.16: 74636588-74636588 on Build GRCh38
rs7191921 Chr.16: 74636483-74636483 on Build GRCh38
rs767312037 Chr.16: 74636521-74636521 on Build GRCh38
rs889152263 Chr.16: 74636565-74636565 on Build GRCh38
rs7190555 Chr.16: 74636508-74636508 on Build GRCh38
rs540343029 Chr.16: 74636610-74636610 on Build GRCh38
rs774078573 Chr.16: 74636618-74636618 on Build GRCh38
rs777224972 Chr.16: 74636484-74636484 on Build GRCh38
rs939878092 Chr.16: 74636517-74636517 on Build GRCh38
rs4888262 Chr.16: 74636560-74636560 on Build GRCh38
rs748923851 Chr.16: 74636598-74636598 on Build GRCh38
rs544324816 Chr.16: 74636519-74636519 on Build GRCh38
rs758670822 Chr.16: 74636555-74636555 on Build GRCh38
rs149000471 Chr.16: 74636538-74636538 on Build GRCh38
rs781492975 Chr.16: 74636580-74636580 on Build GRCh38
rs933873005 Chr.16: 74636547-74636547 on Build GRCh38
rs374480360 Chr.16: 74636531-74636531 on Build GRCh38
rs765527820 Chr.16: 74636561-74636562 on Build GRCh38
rs776051785 Chr.16: 74636591-74636591 on Build GRCh38
rs775211294 Chr.16: 74636512-74636512 on Build GRCh38
rs371423726 Chr.16: 74636583-74636583 on Build GRCh38
rs763643624 Chr.16: 74636518-74636518 on Build GRCh38
rs566050457 Chr.16: 74636552-74636552 on Build GRCh38
rs770275165 Chr.16: 74636590-74636590 on Build GRCh38
rs766228902 Chr.16: 74636549-74636549 on Build GRCh38
rs535860664 Chr.16: 74636677-74636677 on Build GRCh38
rs762757836 Chr.16: 74636514-74636514 on Build GRCh38
rs757378855 Chr.16: 74636577-74636577 on Build GRCh38

View Full Product Details