Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 38599868-38600115 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC105372397
Entrez Gene ID
105372397
Gene Name
uncharacterized LOC105372397
Gene Aliases
-
UniGene
Hs.570011
Gene Chromosome Location
Chr.19: 38596347-38601694 on Build GRCh38
Transcripts
NR_134907.1 NM_001042600.2 NM_007181.5 XM_011526403.1 XM_011526404.1 XM_017026231.1

Gene Symbol
MAP4K1
Entrez Gene ID
11184
Gene Name
mitogen-activated protein kinase kinase kinase kinase 1
Gene Aliases
HPK1
UniGene
Hs.95424
Gene Chromosome Location
Chr.19: 38587640-38618035 on Build GRCh38
Transcripts
NR_134907.1 NM_001042600.2 NM_007181.5 XM_011526403.1 XM_011526404.1 XM_017026231.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7306198 c.1633G>A p.V545M Chr.19: 38599961-38599961 on Build GRCh38
4529342 c.1603G>A p.E535K Chr.19: 38600082-38600082 on Build GRCh38
1750873 c.1594G>A p.A532T Chr.19: 38600091-38600091 on Build GRCh38
5468023 c.1575G>T p.L525L Chr.19: 38600110-38600110 on Build GRCh38
3388962 c.1667C>G p.S556* Chr.19: 38599927-38599927 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111468149 Chr.19: 38600076-38600076 on Build GRCh38
rs969527511 Chr.19: 38600064-38600064 on Build GRCh38
rs745450743 Chr.19: 38599991-38599991 on Build GRCh38
rs753935004 Chr.19: 38600030-38600030 on Build GRCh38
rs949107421 Chr.19: 38599906-38599906 on Build GRCh38
rs111732636 Chr.19: 38600047-38600047 on Build GRCh38
rs138298484 Chr.19: 38600039-38600039 on Build GRCh38
rs202155361 Chr.19: 38600087-38600087 on Build GRCh38
rs200403622 Chr.19: 38599999-38599999 on Build GRCh38
rs760935438 Chr.19: 38600019-38600019 on Build GRCh38
rs758166368 Chr.19: 38599948-38599948 on Build GRCh38
rs150439692 Chr.19: 38600032-38600032 on Build GRCh38
rs770505238 Chr.19: 38599973-38599973 on Build GRCh38
rs377546957 Chr.19: 38600038-38600038 on Build GRCh38
rs369462847 Chr.19: 38599997-38599997 on Build GRCh38
rs754958948 Chr.19: 38599921-38599921 on Build GRCh38
rs780863421 Chr.19: 38600049-38600049 on Build GRCh38
rs754026104 Chr.19: 38599891-38599891 on Build GRCh38
rs888925424 Chr.19: 38599909-38599909 on Build GRCh38
rs780668984 Chr.19: 38599974-38599974 on Build GRCh38
rs777464356 Chr.19: 38599954-38599954 on Build GRCh38
rs772511208 Chr.19: 38600086-38600086 on Build GRCh38
rs140424033 Chr.19: 38599944-38599944 on Build GRCh38
rs748628065 Chr.19: 38600074-38600074 on Build GRCh38
rs192388181 Chr.19: 38599950-38599950 on Build GRCh38
rs531860758 Chr.19: 38599943-38599943 on Build GRCh38
rs372829550 Chr.19: 38599965-38599965 on Build GRCh38
rs766097630 Chr.19: 38600051-38600052 on Build GRCh38
rs376486408 Chr.19: 38599995-38599995 on Build GRCh38
rs771252231 Chr.19: 38600089-38600089 on Build GRCh38
rs773460106 Chr.19: 38600013-38600013 on Build GRCh38
rs890524166 Chr.19: 38600095-38600095 on Build GRCh38
rs772412953 Chr.19: 38600004-38600004 on Build GRCh38
rs756930774 Chr.19: 38600043-38600043 on Build GRCh38
rs533929376 Chr.19: 38599889-38599889 on Build GRCh38
rs113802411 Chr.19: 38600075-38600075 on Build GRCh38
rs373166252 Chr.19: 38600040-38600040 on Build GRCh38
rs917639527 Chr.19: 38599892-38599892 on Build GRCh38
rs766574909 Chr.19: 38600029-38600029 on Build GRCh38
rs745360885 Chr.19: 38600061-38600061 on Build GRCh38
rs41274334 Chr.19: 38600062-38600062 on Build GRCh38
rs373933759 Chr.19: 38600033-38600033 on Build GRCh38
rs779543773 Chr.19: 38600071-38600071 on Build GRCh38

View Full Product Details