Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63738336-63738603 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LIME1
Entrez Gene ID
54923
Gene Name
Lck interacting transmembrane adaptor 1
Gene Aliases
LIME, dJ583P15.4
UniGene
Hs.233220
Gene Chromosome Location
Chr.20: 63735701-63739107 on Build GRCh38
Transcripts
NM_001305654.1 NM_001305655.1 NM_017806.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6424848 c.441C>A p.N147K Chr.20: 63738355-63738355 on Build GRCh38
5564685 c.505C>T p.R169C Chr.20: 63738419-63738419 on Build GRCh38
6215661 c.460C>T p.P154S Chr.20: 63738374-63738374 on Build GRCh38
1197288 c.464G>C p.G155A Chr.20: 63738378-63738378 on Build GRCh38
5366126 c.518G>A p.R173H Chr.20: 63738432-63738432 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs971974937 Chr.20: 63738399-63738399 on Build GRCh38
rs781438076 Chr.20: 63738486-63738486 on Build GRCh38
rs766064783 Chr.20: 63738361-63738361 on Build GRCh38
rs572812489 Chr.20: 63738566-63738566 on Build GRCh38
rs749836196 Chr.20: 63738583-63738583 on Build GRCh38
rs773924642 Chr.20: 63738530-63738530 on Build GRCh38
rs990139068 Chr.20: 63738460-63738460 on Build GRCh38
rs760658107 Chr.20: 63738537-63738537 on Build GRCh38
rs752190844 Chr.20: 63738534-63738534 on Build GRCh38
rs751232208 Chr.20: 63738367-63738367 on Build GRCh38
rs773510884 Chr.20: 63738356-63738356 on Build GRCh38
rs752620750 Chr.20: 63738396-63738396 on Build GRCh38
rs571836738 Chr.20: 63738515-63738515 on Build GRCh38
rs977468123 Chr.20: 63738400-63738400 on Build GRCh38
rs747267914 Chr.20: 63738478-63738478 on Build GRCh38
rs780375335 Chr.20: 63738459-63738459 on Build GRCh38
rs768791027 Chr.20: 63738479-63738479 on Build GRCh38
rs188602790 Chr.20: 63738504-63738504 on Build GRCh38
rs1151624 Chr.20: 63738542-63738542 on Build GRCh38
rs181321157 Chr.20: 63738544-63738544 on Build GRCh38
rs373042264 Chr.20: 63738386-63738386 on Build GRCh38
rs766999697 Chr.20: 63738532-63738532 on Build GRCh38
rs763995300 Chr.20: 63738538-63738538 on Build GRCh38
rs771312330 Chr.20: 63738525-63738525 on Build GRCh38
rs777618606 Chr.20: 63738432-63738432 on Build GRCh38
rs748678885 Chr.20: 63738505-63738505 on Build GRCh38
rs771960419 Chr.20: 63738486-63738486 on Build GRCh38
rs753827588 Chr.20: 63738541-63738541 on Build GRCh38
rs964422221 Chr.20: 63738483-63738483 on Build GRCh38
rs756062890 Chr.20: 63738417-63738417 on Build GRCh38
rs780856863 Chr.20: 63738522-63738522 on Build GRCh38
rs755296605 Chr.20: 63738545-63738545 on Build GRCh38
rs765153498 Chr.20: 63738543-63738543 on Build GRCh38
rs758625007 Chr.20: 63738455-63738455 on Build GRCh38
rs1011286125 Chr.20: 63738407-63738407 on Build GRCh38
rs866562108 Chr.20: 63738489-63738489 on Build GRCh38
rs1017667308 Chr.20: 63738480-63738480 on Build GRCh38
rs1031751351 Chr.20: 63738409-63738409 on Build GRCh38
rs781297574 Chr.20: 63738550-63738550 on Build GRCh38
rs370081076 Chr.20: 63738576-63738576 on Build GRCh38
rs376105385 Chr.20: 63738570-63738570 on Build GRCh38
rs770274505 Chr.20: 63738506-63738506 on Build GRCh38
rs763175336 Chr.20: 63738518-63738518 on Build GRCh38
rs377345131 Chr.20: 63738446-63738446 on Build GRCh38
rs759004080 Chr.20: 63738378-63738378 on Build GRCh38
rs768953868 Chr.20: 63738584-63738584 on Build GRCh38

View Full Product Details