Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 153390176-153390443 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
S100A8
Entrez Gene ID
6279
Gene Name
S100 calcium binding protein A8
Gene Aliases
60B8AG, CAGA, CFAG, CGLA, CP-10, L1Ag, MA387, MIF, MRP8, NIF, P8
UniGene
Hs.416073
Gene Chromosome Location
Chr.1: 153390032-153422583 on Build GRCh38
Transcripts
NM_001319196.1 NM_001319197.1 NM_001319198.1 NM_001319201.1 NM_002964.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6274255 c.183C>A p.N61K Chr.1: 153390202-153390202 on Build GRCh38
4944968 c.168A>T p.K56N Chr.1: 153390217-153390217 on Build GRCh38
4962494 c.185C>G p.T62S Chr.1: 153390200-153390200 on Build GRCh38
6019548 c.121G>C p.E41Q Chr.1: 153390415-153390415 on Build GRCh38
360700 c.146A>G p.K49R Chr.1: 153390239-153390239 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746791160 Chr.1: 153390373-153390373 on Build GRCh38
rs979978083 Chr.1: 153390399-153390399 on Build GRCh38
rs878879901 Chr.1: 153390390-153390390 on Build GRCh38
rs375159557 Chr.1: 153390380-153390380 on Build GRCh38
rs759829878 Chr.1: 153390417-153390417 on Build GRCh38
rs751883285 Chr.1: 153390416-153390416 on Build GRCh38
rs761231293 Chr.1: 153390290-153390290 on Build GRCh38
rs1035912446 Chr.1: 153390341-153390341 on Build GRCh38
rs756182092 Chr.1: 153390361-153390361 on Build GRCh38
rs768285127 Chr.1: 153390374-153390374 on Build GRCh38
rs772018841 Chr.1: 153390260-153390260 on Build GRCh38
rs758791841 Chr.1: 153390206-153390206 on Build GRCh38
rs879122501 Chr.1: 153390392-153390392 on Build GRCh38
rs776146680 Chr.1: 153390286-153390286 on Build GRCh38
rs879072116 Chr.1: 153390245-153390245 on Build GRCh38
rs41264638 Chr.1: 153390312-153390312 on Build GRCh38
rs879048827 Chr.1: 153390393-153390393 on Build GRCh38
rs192475448 Chr.1: 153390291-153390291 on Build GRCh38
rs1050734822 Chr.1: 153390285-153390285 on Build GRCh38
rs534423176 Chr.1: 153390332-153390332 on Build GRCh38
rs547659803 Chr.1: 153390333-153390333 on Build GRCh38
rs769263523 Chr.1: 153390389-153390389 on Build GRCh38
rs376188722 Chr.1: 153390354-153390354 on Build GRCh38
rs768248544 Chr.1: 153390271-153390271 on Build GRCh38
rs748236363 Chr.1: 153390228-153390228 on Build GRCh38
rs1035822446 Chr.1: 153390367-153390367 on Build GRCh38
rs780199591 Chr.1: 153390212-153390212 on Build GRCh38
rs998675392 Chr.1: 153390325-153390325 on Build GRCh38
rs879000199 Chr.1: 153390246-153390246 on Build GRCh38
rs779001718 Chr.1: 153390237-153390237 on Build GRCh38
rs761347222 Chr.1: 153390385-153390385 on Build GRCh38
rs199736559 Chr.1: 153390348-153390348 on Build GRCh38
rs139987242 Chr.1: 153390418-153390418 on Build GRCh38
rs755195830 Chr.1: 153390224-153390224 on Build GRCh38
rs775461109 Chr.1: 153390261-153390261 on Build GRCh38
rs752757891 Chr.1: 153390360-153390360 on Build GRCh38
rs763499598 Chr.1: 153390295-153390295 on Build GRCh38
rs750949807 Chr.1: 153390292-153390292 on Build GRCh38
rs763436258 Chr.1: 153390411-153390411 on Build GRCh38
rs760218947 Chr.1: 153390307-153390307 on Build GRCh38
rs888592743 Chr.1: 153390296-153390296 on Build GRCh38
rs747209514 Chr.1: 153390213-153390213 on Build GRCh38
rs755215366 Chr.1: 153390349-153390349 on Build GRCh38
rs879172337 Chr.1: 153390394-153390395 on Build GRCh38
rs377695571 Chr.1: 153390365-153390365 on Build GRCh38
rs80122691 Chr.1: 153390323-153390323 on Build GRCh38
rs1043089043 Chr.1: 153390317-153390317 on Build GRCh38
rs772758212 Chr.1: 153390396-153390396 on Build GRCh38
rs878891089 Chr.1: 153390388-153390388 on Build GRCh38
rs766699265 Chr.1: 153390344-153390344 on Build GRCh38
rs781328492 Chr.1: 153390227-153390227 on Build GRCh38
rs879236587 Chr.1: 153390247-153390247 on Build GRCh38
rs1013240106 Chr.1: 153390368-153390368 on Build GRCh38
rs766936688 Chr.1: 153390415-153390415 on Build GRCh38
rs745782999 Chr.1: 153390370-153390370 on Build GRCh38
rs879250129 Chr.1: 153390248-153390248 on Build GRCh38
rs147024535 Chr.1: 153390372-153390372 on Build GRCh38
rs770846885 Chr.1: 153390229-153390229 on Build GRCh38
rs113251289 Chr.1: 153390394-153390394 on Build GRCh38
rs771726018 Chr.1: 153390270-153390270 on Build GRCh38
rs933557072 Chr.1: 153390283-153390283 on Build GRCh38

View Full Product Details