Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 15426238-15426469 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
METTL6
Entrez Gene ID
131965
Gene Name
methyltransferase like 6
Gene Aliases
-
UniGene
Hs.149487
Gene Chromosome Location
Chr.3: 15381275-15427580 on Build GRCh38
Transcripts
NM_001301790.1 NM_001301791.1 NM_001301792.1 NM_152396.3 XM_005264867.3 XM_005264870.4 XM_006712970.3 XM_006712972.3 XM_011533357.2 XM_017005718.1 XM_017005719.1 XM_017005720.1 XM_017005721.1 XM_017005722.1 XM_017005723.1 XM_017005724.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4866514 c.168C>A p.F56L Chr.3: 15426344-15426344 on Build GRCh38
4866981 c.196G>A p.E66K Chr.3: 15426316-15426316 on Build GRCh38
261984 c.168C>A p.F56L Chr.3: 15426344-15426344 on Build GRCh38
479610 c.225G>T p.E75D Chr.3: 15426287-15426287 on Build GRCh38
5430101 c.202G>A p.E68K Chr.3: 15426310-15426310 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376995362 Chr.3: 15426269-15426269 on Build GRCh38
rs777439469 Chr.3: 15426330-15426330 on Build GRCh38
rs372690037 Chr.3: 15426336-15426336 on Build GRCh38
rs373785631 Chr.3: 15426419-15426419 on Build GRCh38
rs61734701 Chr.3: 15426351-15426351 on Build GRCh38
rs1012840776 Chr.3: 15426404-15426404 on Build GRCh38
rs1004149798 Chr.3: 15426344-15426344 on Build GRCh38
rs767540189 Chr.3: 15426412-15426412 on Build GRCh38
rs749611723 Chr.3: 15426331-15426331 on Build GRCh38
rs79200789 Chr.3: 15426341-15426341 on Build GRCh38
rs775423616 Chr.3: 15426360-15426360 on Build GRCh38
rs766088421 Chr.3: 15426395-15426395 on Build GRCh38
rs754442228 Chr.3: 15426295-15426295 on Build GRCh38
rs777028015 Chr.3: 15426391-15426391 on Build GRCh38
rs566848011 Chr.3: 15426377-15426377 on Build GRCh38
rs377494656 Chr.3: 15426439-15426439 on Build GRCh38
rs535856188 Chr.3: 15426290-15426290 on Build GRCh38
rs892029686 Chr.3: 15426289-15426289 on Build GRCh38
rs777354223 Chr.3: 15426420-15426420 on Build GRCh38
rs771220576 Chr.3: 15426335-15426335 on Build GRCh38
rs780561920 Chr.3: 15426317-15426317 on Build GRCh38
rs567294912 Chr.3: 15426271-15426271 on Build GRCh38
rs768972630 Chr.3: 15426380-15426380 on Build GRCh38
rs368652669 Chr.3: 15426276-15426276 on Build GRCh38
rs755738163 Chr.3: 15426323-15426323 on Build GRCh38
rs765658558 Chr.3: 15426319-15426321 on Build GRCh38
rs751435093 Chr.3: 15426403-15426403 on Build GRCh38
rs758313932 Chr.3: 15426270-15426270 on Build GRCh38
rs752171481 Chr.3: 15426414-15426414 on Build GRCh38
rs759373863 Chr.3: 15426410-15426410 on Build GRCh38
rs750779510 Chr.3: 15426366-15426366 on Build GRCh38
rs931371915 Chr.3: 15426313-15426313 on Build GRCh38
rs960052795 Chr.3: 15426398-15426398 on Build GRCh38
rs370811487 Chr.3: 15426357-15426357 on Build GRCh38
rs752261742 Chr.3: 15426319-15426319 on Build GRCh38
rs762776430 Chr.3: 15426394-15426394 on Build GRCh38
rs761887045 Chr.3: 15426315-15426319 on Build GRCh38
rs760978562 Chr.3: 15426272-15426272 on Build GRCh38
rs760889969 Chr.3: 15426363-15426363 on Build GRCh38
rs751464134 Chr.3: 15426291-15426291 on Build GRCh38
rs551800321 Chr.3: 15426287-15426287 on Build GRCh38

View Full Product Details