Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 5788540-5788787 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR52N1
Entrez Gene ID
79473
Gene Name
olfactory receptor family 52 subfamily N member 1
Gene Aliases
OR11-61
UniGene
Hs.690209
Gene Chromosome Location
Chr.11: 5787854-5788816 on Build GRCh38
Transcripts
NM_001001913.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1260401 c.133T>G p.F45V Chr.11: 5788684-5788684 on Build GRCh38
7218138 c.57C>T p.I19I Chr.11: 5788760-5788760 on Build GRCh38
1286869 c.59C>A p.P20H Chr.11: 5788758-5788758 on Build GRCh38
6069447 c.271C>G p.L91V Chr.11: 5788546-5788546 on Build GRCh38
1604792 c.257T>C p.I86T Chr.11: 5788560-5788560 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs868503228 Chr.11: 5788575-5788576 on Build GRCh38
rs761714208 Chr.11: 5788678-5788678 on Build GRCh38
rs774981754 Chr.11: 5788655-5788655 on Build GRCh38
rs768454072 Chr.11: 5788664-5788664 on Build GRCh38
rs745363471 Chr.11: 5788650-5788650 on Build GRCh38
rs970950324 Chr.11: 5788744-5788744 on Build GRCh38
rs778109488 Chr.11: 5788721-5788721 on Build GRCh38
rs201569028 Chr.11: 5788717-5788717 on Build GRCh38
rs746610022 Chr.11: 5788656-5788656 on Build GRCh38
rs12365487 Chr.11: 5788581-5788581 on Build GRCh38
rs370553004 Chr.11: 5788758-5788758 on Build GRCh38
rs778398396 Chr.11: 5788638-5788638 on Build GRCh38
rs751823446 Chr.11: 5788589-5788589 on Build GRCh38
rs549551174 Chr.11: 5788602-5788602 on Build GRCh38
rs759719361 Chr.11: 5788703-5788703 on Build GRCh38
rs151102157 Chr.11: 5788749-5788749 on Build GRCh38
rs997419773 Chr.11: 5788632-5788632 on Build GRCh38
rs149066849 Chr.11: 5788636-5788636 on Build GRCh38
rs539044197 Chr.11: 5788714-5788714 on Build GRCh38
rs1000706209 Chr.11: 5788665-5788665 on Build GRCh38
rs746522145 Chr.11: 5788756-5788756 on Build GRCh38
rs374600480 Chr.11: 5788710-5788710 on Build GRCh38
rs142442713 Chr.11: 5788577-5788577 on Build GRCh38
rs767735888 Chr.11: 5788704-5788704 on Build GRCh38
rs781492158 Chr.11: 5788625-5788625 on Build GRCh38
rs557902606 Chr.11: 5788723-5788723 on Build GRCh38
rs766532712 Chr.11: 5788696-5788696 on Build GRCh38
rs570634505 Chr.11: 5788682-5788682 on Build GRCh38
rs143758698 Chr.11: 5788671-5788671 on Build GRCh38
rs147594592 Chr.11: 5788594-5788594 on Build GRCh38
rs868019418 Chr.11: 5788666-5788666 on Build GRCh38
rs1036212485 Chr.11: 5788590-5788590 on Build GRCh38
rs748512030 Chr.11: 5788635-5788635 on Build GRCh38
rs773268875 Chr.11: 5788692-5788692 on Build GRCh38
rs750412950 Chr.11: 5788571-5788571 on Build GRCh38
rs11039038 Chr.11: 5788760-5788760 on Build GRCh38
rs751664250 Chr.11: 5788702-5788702 on Build GRCh38
rs760388282 Chr.11: 5788612-5788612 on Build GRCh38
rs1056532818 Chr.11: 5788591-5788591 on Build GRCh38
rs758565262 Chr.11: 5788579-5788579 on Build GRCh38
rs762913243 Chr.11: 5788693-5788693 on Build GRCh38
rs757726223 Chr.11: 5788741-5788741 on Build GRCh38

View Full Product Details