Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 2931144-2931389 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MFSD10
Entrez Gene ID
10227
Gene Name
major facilitator superfamily domain containing 10
Gene Aliases
TETRAN, TETTRAN
UniGene
Hs.632581
Gene Chromosome Location
Chr.4: 2930560-2934859 on Build GRCh38
Transcripts
NM_001120.4 NM_001146069.1 XM_005247927.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7282816 c.1153G>A p.A385T Chr.4: 2931236-2931236 on Build GRCh38
6252493 c.1146C>A p.S382S Chr.4: 2931243-2931243 on Build GRCh38
3428459 c.1120G>A p.A374T Chr.4: 2931269-2931269 on Build GRCh38
3714864 c.1145C>G p.S382C Chr.4: 2931244-2931244 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs199793785 Chr.4: 2931249-2931249 on Build GRCh38
rs551811310 Chr.4: 2931236-2931236 on Build GRCh38
rs992725050 Chr.4: 2931239-2931239 on Build GRCh38
rs868552880 Chr.4: 2931253-2931253 on Build GRCh38
rs749300630 Chr.4: 2931345-2931345 on Build GRCh38
rs1025534564 Chr.4: 2931313-2931313 on Build GRCh38
rs766906371 Chr.4: 2931316-2931316 on Build GRCh38
rs750285503 Chr.4: 2931264-2931265 on Build GRCh38
rs201884555 Chr.4: 2931300-2931300 on Build GRCh38
rs747282222 Chr.4: 2931243-2931243 on Build GRCh38
rs55754437 Chr.4: 2931304-2931304 on Build GRCh38
rs374316576 Chr.4: 2931191-2931191 on Build GRCh38
rs751967558 Chr.4: 2931331-2931331 on Build GRCh38
rs368892106 Chr.4: 2931332-2931332 on Build GRCh38
rs777687146 Chr.4: 2931343-2931343 on Build GRCh38
rs370120563 Chr.4: 2931205-2931205 on Build GRCh38
rs921171036 Chr.4: 2931267-2931267 on Build GRCh38
rs79362137 Chr.4: 2931192-2931192 on Build GRCh38
rs534014752 Chr.4: 2931244-2931244 on Build GRCh38
rs917030215 Chr.4: 2931240-2931240 on Build GRCh38
rs879852373 Chr.4: 2931336-2931336 on Build GRCh38
rs770756210 Chr.4: 2931289-2931289 on Build GRCh38
rs372410385 Chr.4: 2931351-2931351 on Build GRCh38
rs796113972 Chr.4: 2931315-2931315 on Build GRCh38
rs374231622 Chr.4: 2931206-2931206 on Build GRCh38
rs772887612 Chr.4: 2931176-2931176 on Build GRCh38
rs576820601 Chr.4: 2931295-2931295 on Build GRCh38
rs115907605 Chr.4: 2931209-2931209 on Build GRCh38
rs778546621 Chr.4: 2931164-2931165 on Build GRCh38
rs959872760 Chr.4: 2931306-2931306 on Build GRCh38
rs1013771758 Chr.4: 2931307-2931307 on Build GRCh38
rs758291800 Chr.4: 2931310-2931311 on Build GRCh38
rs940132085 Chr.4: 2931242-2931242 on Build GRCh38
rs375830658 Chr.4: 2931362-2931362 on Build GRCh38
rs868104271 Chr.4: 2931261-2931261 on Build GRCh38
rs758948732 Chr.4: 2931312-2931312 on Build GRCh38
rs767157020 Chr.4: 2931296-2931296 on Build GRCh38
rs925002074 Chr.4: 2931189-2931189 on Build GRCh38
rs752743120 Chr.4: 2931340-2931340 on Build GRCh38
rs553798078 Chr.4: 2931272-2931272 on Build GRCh38
rs781487416 Chr.4: 2931266-2931266 on Build GRCh38
rs1032073320 Chr.4: 2931301-2931301 on Build GRCh38
rs374126490 Chr.4: 2931270-2931270 on Build GRCh38
rs866046500 Chr.4: 2931260-2931260 on Build GRCh38
rs749070343 Chr.4: 2931273-2931273 on Build GRCh38
rs200589034 Chr.4: 2931234-2931234 on Build GRCh38
rs908535097 Chr.4: 2931224-2931224 on Build GRCh38
rs781507727 Chr.4: 2931334-2931334 on Build GRCh38
rs750772949 Chr.4: 2931218-2931218 on Build GRCh38
rs748192658 Chr.4: 2931269-2931269 on Build GRCh38
rs1008143909 Chr.4: 2931278-2931278 on Build GRCh38
rs370712161 Chr.4: 2931210-2931210 on Build GRCh38
rs756181389 Chr.4: 2931342-2931342 on Build GRCh38
rs368189249 Chr.4: 2931290-2931290 on Build GRCh38
rs201620380 Chr.4: 2931170-2931170 on Build GRCh38
rs144539170 Chr.4: 2931184-2931184 on Build GRCh38
rs1036998634 Chr.4: 2931292-2931292 on Build GRCh38
rs781446411 Chr.4: 2931317-2931317 on Build GRCh38
rs780532870 Chr.4: 2931237-2931237 on Build GRCh38
rs144879290 Chr.4: 2931238-2931238 on Build GRCh38
rs750976459 Chr.4: 2931305-2931305 on Build GRCh38
rs1015522842 Chr.4: 2931197-2931197 on Build GRCh38
rs371940381 Chr.4: 2931302-2931302 on Build GRCh38
rs977129546 Chr.4: 2931346-2931346 on Build GRCh38
rs762752995 Chr.4: 2931177-2931177 on Build GRCh38

View Full Product Details