Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 46825353-46825578 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MEP1A
Entrez Gene ID
4224
Gene Name
meprin A subunit alpha
Gene Aliases
PPHA
UniGene
Hs.179704
Gene Chromosome Location
Chr.6: 46793357-46845987 on Build GRCh38
Transcripts
NM_005588.2 XM_011514628.1 XM_011514629.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3830681 c.696C>A p.I232I Chr.6: 46825411-46825411 on Build GRCh38
402786 c.717C>T p.I239I Chr.6: 46825432-46825432 on Build GRCh38
6193306 c.745G>A p.D249N Chr.6: 46825460-46825460 on Build GRCh38
6865205 c.749T>C p.L250S Chr.6: 46825464-46825464 on Build GRCh38
4844223 c.753G>C p.E251D Chr.6: 46825468-46825468 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773986621 Chr.6: 46825444-46825444 on Build GRCh38
rs747199369 Chr.6: 46825502-46825502 on Build GRCh38
rs775825864 Chr.6: 46825427-46825427 on Build GRCh38
rs755976475 Chr.6: 46825404-46825404 on Build GRCh38
rs773894954 Chr.6: 46825540-46825540 on Build GRCh38
rs769018940 Chr.6: 46825433-46825433 on Build GRCh38
rs781616698 Chr.6: 46825516-46825516 on Build GRCh38
rs200557738 Chr.6: 46825405-46825405 on Build GRCh38
rs149650040 Chr.6: 46825414-46825414 on Build GRCh38
rs752353235 Chr.6: 46825461-46825461 on Build GRCh38
rs778875477 Chr.6: 46825479-46825479 on Build GRCh38
rs768767681 Chr.6: 46825512-46825512 on Build GRCh38
rs201836483 Chr.6: 46825426-46825426 on Build GRCh38
rs146158038 Chr.6: 46825432-46825432 on Build GRCh38
rs773102061 Chr.6: 46825483-46825483 on Build GRCh38
rs1030396152 Chr.6: 46825464-46825464 on Build GRCh38
rs199863964 Chr.6: 46825420-46825420 on Build GRCh38
rs368226667 Chr.6: 46825529-46825529 on Build GRCh38
rs148650252 Chr.6: 46825492-46825492 on Build GRCh38
rs752411870 Chr.6: 46825402-46825402 on Build GRCh38
rs748582278 Chr.6: 46825522-46825522 on Build GRCh38
rs775145526 Chr.6: 46825428-46825429 on Build GRCh38
rs751119683 Chr.6: 46825388-46825388 on Build GRCh38
rs138133276 Chr.6: 46825407-46825407 on Build GRCh38
rs370570724 Chr.6: 46825542-46825542 on Build GRCh38
rs758436819 Chr.6: 46825487-46825487 on Build GRCh38
rs150837750 Chr.6: 46825394-46825394 on Build GRCh38
rs763889686 Chr.6: 46825470-46825470 on Build GRCh38
rs757197749 Chr.6: 46825416-46825416 on Build GRCh38
rs368922958 Chr.6: 46825478-46825478 on Build GRCh38
rs755741706 Chr.6: 46825466-46825466 on Build GRCh38
rs759167108 Chr.6: 46825447-46825447 on Build GRCh38
rs767337393 Chr.6: 46825401-46825401 on Build GRCh38
rs762345708 Chr.6: 46825439-46825439 on Build GRCh38
rs954778640 Chr.6: 46825468-46825468 on Build GRCh38
rs6920863 Chr.6: 46825438-46825438 on Build GRCh38
rs143801305 Chr.6: 46825455-46825455 on Build GRCh38
rs566847239 Chr.6: 46825440-46825440 on Build GRCh38
rs146350772 Chr.6: 46825485-46825485 on Build GRCh38

View Full Product Details