Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 30466329-30466564 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEC14L3
Entrez Gene ID
266629
Gene Name
SEC14 like lipid binding 3
Gene Aliases
TAP2
UniGene
Hs.505601
Gene Chromosome Location
Chr.22: 30447959-30472047 on Build GRCh38
Transcripts
NM_001257378.1 NM_001257379.1 NM_001257382.1 NM_174975.4 XM_011530128.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4971775 c.520-5C>T p.? Chr.22: 30466399-30466399 on Build GRCh38
1714270 c.532C>T p.L178F Chr.22: 30466382-30466382 on Build GRCh38
4298611 c.379C>A p.L127M Chr.22: 30466358-30466358 on Build GRCh38
4869121 c.359A>G p.E120G Chr.22: 30466378-30466378 on Build GRCh38
1033285 c.536A>G p.E179G Chr.22: 30466378-30466378 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1016611652 Chr.22: 30466465-30466465 on Build GRCh38
rs969037827 Chr.22: 30466474-30466474 on Build GRCh38
rs980393377 Chr.22: 30466477-30466477 on Build GRCh38
rs772482076 Chr.22: 30466415-30466415 on Build GRCh38
rs541632016 Chr.22: 30466512-30466512 on Build GRCh38
rs1025871465 Chr.22: 30466419-30466419 on Build GRCh38
rs377536593 Chr.22: 30466400-30466400 on Build GRCh38
rs773763074 Chr.22: 30466430-30466430 on Build GRCh38
rs747510552 Chr.22: 30466440-30466440 on Build GRCh38
rs779210950 Chr.22: 30466403-30466403 on Build GRCh38
rs756826492 Chr.22: 30466449-30466449 on Build GRCh38
rs752568506 Chr.22: 30466360-30466360 on Build GRCh38
rs1024761778 Chr.22: 30466460-30466460 on Build GRCh38
rs771020915 Chr.22: 30466441-30466441 on Build GRCh38
rs1013325709 Chr.22: 30466442-30466442 on Build GRCh38
rs1032012886 Chr.22: 30466503-30466503 on Build GRCh38
rs116469787 Chr.22: 30466386-30466386 on Build GRCh38
rs751513216 Chr.22: 30466376-30466376 on Build GRCh38
rs576812816 Chr.22: 30466362-30466362 on Build GRCh38
rs756791332 Chr.22: 30466385-30466385 on Build GRCh38
rs745512625 Chr.22: 30466393-30466393 on Build GRCh38
rs1057995 Chr.22: 30466413-30466413 on Build GRCh38
rs540971001 Chr.22: 30466374-30466374 on Build GRCh38
rs560091798 Chr.22: 30466395-30466395 on Build GRCh38
rs866721709 Chr.22: 30466423-30466423 on Build GRCh38
rs765071820 Chr.22: 30466357-30466357 on Build GRCh38

View Full Product Details