Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 78469164-78469401 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NARS2
Entrez Gene ID
79731
Gene Name
asparaginyl-tRNA synthetase 2, mitochondrial (putative)
Gene Aliases
DFNB94, SLM5, asnRS
UniGene
Hs.503389
Gene Chromosome Location
Chr.11: 78435961-78574864 on Build GRCh38
Transcripts
NM_001243251.1 NM_024678.5 XM_011545253.1 XM_017018303.1 XM_017018304.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7421067 c.1024G>A p.E342K Chr.11: 78469249-78469249 on Build GRCh38
6721980 c.1014C>T p.T338T Chr.11: 78469259-78469259 on Build GRCh38
4705508 c.990A>G p.L330L Chr.11: 78469283-78469283 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs377264119 Chr.11: 78469206-78469206 on Build GRCh38
rs777355564 Chr.11: 78469350-78469350 on Build GRCh38
rs746714131 Chr.11: 78469352-78469352 on Build GRCh38
rs544222321 Chr.11: 78469349-78469349 on Build GRCh38
rs779756829 Chr.11: 78469218-78469218 on Build GRCh38
rs764473733 Chr.11: 78469275-78469275 on Build GRCh38
rs981811147 Chr.11: 78469338-78469338 on Build GRCh38
rs753555379 Chr.11: 78469312-78469312 on Build GRCh38
rs781422969 Chr.11: 78469205-78469205 on Build GRCh38
rs756323353 Chr.11: 78469298-78469298 on Build GRCh38
rs111319410 Chr.11: 78469263-78469263 on Build GRCh38
rs749315308 Chr.11: 78469222-78469222 on Build GRCh38
rs544972589 Chr.11: 78469283-78469283 on Build GRCh38
rs766342345 Chr.11: 78469291-78469291 on Build GRCh38
rs376091227 Chr.11: 78469237-78469237 on Build GRCh38
rs112414788 Chr.11: 78469213-78469213 on Build GRCh38
rs775980355 Chr.11: 78469264-78469264 on Build GRCh38
rs747580509 Chr.11: 78469236-78469236 on Build GRCh38
rs775958836 Chr.11: 78469365-78469365 on Build GRCh38
rs903245886 Chr.11: 78469214-78469214 on Build GRCh38
rs781047471 Chr.11: 78469353-78469353 on Build GRCh38
rs754983949 Chr.11: 78469316-78469316 on Build GRCh38
rs760348257 Chr.11: 78469255-78469255 on Build GRCh38
rs754579488 Chr.11: 78469277-78469281 on Build GRCh38
rs147815304 Chr.11: 78469226-78469226 on Build GRCh38
rs565224393 Chr.11: 78469304-78469304 on Build GRCh38
rs768467263 Chr.11: 78469224-78469224 on Build GRCh38
rs770289829 Chr.11: 78469260-78469260 on Build GRCh38
rs751118197 Chr.11: 78469212-78469213 on Build GRCh38
rs184653643 Chr.11: 78469322-78469322 on Build GRCh38
rs773025539 Chr.11: 78469249-78469249 on Build GRCh38
rs377435131 Chr.11: 78469287-78469287 on Build GRCh38
rs760946077 Chr.11: 78469308-78469308 on Build GRCh38
rs748073776 Chr.11: 78469335-78469335 on Build GRCh38
rs937854474 Chr.11: 78469372-78469372 on Build GRCh38
rs750645454 Chr.11: 78469295-78469295 on Build GRCh38
rs368627038 Chr.11: 78469357-78469357 on Build GRCh38
rs950440420 Chr.11: 78469332-78469332 on Build GRCh38
rs752062550 Chr.11: 78469282-78469282 on Build GRCh38
rs200255394 Chr.11: 78469216-78469216 on Build GRCh38
rs34950075 Chr.11: 78469265-78469265 on Build GRCh38

View Full Product Details