Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45489265-45489506 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RTN2
Entrez Gene ID
6253
Gene Name
reticulon 2
Gene Aliases
NSP2, NSPL1, NSPLI, SPG12
UniGene
Hs.47517
Gene Chromosome Location
Chr.19: 45485288-45497055 on Build GRCh38
Transcripts
NM_005619.4 NM_206900.2 NM_206901.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4515633 c.1220_1221GG>AA p.G407E Chr.19: 45489366-45489367 on Build GRCh38
5550855 c.1167C>T p.C389C Chr.19: 45489420-45489420 on Build GRCh38
6410086 c.1228G>T p.A410S Chr.19: 45489359-45489359 on Build GRCh38
76304 c.1192C>T p.R398C Chr.19: 45489395-45489395 on Build GRCh38
4127712 c.1209C>G p.A403A Chr.19: 45489378-45489378 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775924273 Chr.19: 45489349-45489349 on Build GRCh38
rs980714416 Chr.19: 45489288-45489288 on Build GRCh38
rs755919124 Chr.19: 45489406-45489406 on Build GRCh38
rs202061449 Chr.19: 45489453-45489453 on Build GRCh38
rs751685341 Chr.19: 45489469-45489469 on Build GRCh38
rs150034426 Chr.19: 45489389-45489389 on Build GRCh38
rs768120941 Chr.19: 45489343-45489343 on Build GRCh38
rs532500975 Chr.19: 45489452-45489452 on Build GRCh38
rs140427245 Chr.19: 45489378-45489378 on Build GRCh38
rs776793996 Chr.19: 45489445-45489445 on Build GRCh38
rs140494585 Chr.19: 45489487-45489487 on Build GRCh38
rs575595947 Chr.19: 45489412-45489412 on Build GRCh38
rs1039418305 Chr.19: 45489395-45489395 on Build GRCh38
rs781104615 Chr.19: 45489402-45489402 on Build GRCh38
rs777775690 Chr.19: 45489485-45489485 on Build GRCh38
rs146778767 Chr.19: 45489458-45489458 on Build GRCh38
rs45532933 Chr.19: 45489444-45489444 on Build GRCh38
rs143937661 Chr.19: 45489419-45489419 on Build GRCh38
rs752618262 Chr.19: 45489405-45489405 on Build GRCh38
rs773962360 Chr.19: 45489333-45489333 on Build GRCh38
rs978898311 Chr.19: 45489319-45489319 on Build GRCh38
rs771745205 Chr.19: 45489428-45489428 on Build GRCh38
rs776040368 Chr.19: 45489440-45489440 on Build GRCh38
rs761067091 Chr.19: 45489357-45489357 on Build GRCh38
rs1022880283 Chr.19: 45489306-45489306 on Build GRCh38
rs777385348 Chr.19: 45489411-45489411 on Build GRCh38
rs868042088 Chr.19: 45489354-45489354 on Build GRCh38
rs772890773 Chr.19: 45489295-45489295 on Build GRCh38
rs925869493 Chr.19: 45489373-45489373 on Build GRCh38
rs917119159 Chr.19: 45489348-45489348 on Build GRCh38
rs200512597 Chr.19: 45489377-45489377 on Build GRCh38
rs960805012 Chr.19: 45489317-45489317 on Build GRCh38
rs200948085 Chr.19: 45489483-45489483 on Build GRCh38
rs185630222 Chr.19: 45489334-45489334 on Build GRCh38
rs369158895 Chr.19: 45489370-45489370 on Build GRCh38
rs778762660 Chr.19: 45489423-45489423 on Build GRCh38
rs45548637 Chr.19: 45489286-45489286 on Build GRCh38
rs61729959 Chr.19: 45489347-45489347 on Build GRCh38
rs977755323 Chr.19: 45489371-45489371 on Build GRCh38
rs758370815 Chr.19: 45489420-45489420 on Build GRCh38
rs142458832 Chr.19: 45489446-45489446 on Build GRCh38
rs748945582 Chr.19: 45489305-45489305 on Build GRCh38
rs765432276 Chr.19: 45489447-45489447 on Build GRCh38
rs542004944 Chr.19: 45489376-45489376 on Build GRCh38
rs372352187 Chr.19: 45489316-45489316 on Build GRCh38
rs758531242 Chr.19: 45489393-45489393 on Build GRCh38
rs761037026 Chr.19: 45489441-45489441 on Build GRCh38

View Full Product Details