Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 14593922-14594175 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC388813
Entrez Gene ID
388813
Gene Name
uncharacterized protein ENSP00000383407-like
Gene Aliases
-
UniGene
Hs.334177
Gene Chromosome Location
Chr.21: 14591392-14643107 on Build GRCh38
Transcripts
NM_001256579.2 NR_046512.2

Gene Symbol
SAMSN1-AS1
Entrez Gene ID
100874190
Gene Name
SAMSN1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.563053
Gene Chromosome Location
Chr.21: 14582202-14598303 on Build GRCh38
Transcripts
NM_001256579.2 NR_046512.2

SNP Detail

SNP ID Chromosome Location
rs1047447733 Chr.21: 14593988-14593988 on Build GRCh38
rs757513421 Chr.21: 14594050-14594050 on Build GRCh38
rs460852 Chr.21: 14594100-14594100 on Build GRCh38
rs763746148 Chr.21: 14594083-14594083 on Build GRCh38
rs1005809177 Chr.21: 14594091-14594091 on Build GRCh38
rs780583409 Chr.21: 14594102-14594102 on Build GRCh38
rs747479795 Chr.21: 14594105-14594105 on Build GRCh38
rs556550298 Chr.21: 14594026-14594026 on Build GRCh38
rs745990717 Chr.21: 14594073-14594073 on Build GRCh38
rs542968791 Chr.21: 14593970-14593970 on Build GRCh38
rs754861913 Chr.21: 14593992-14593992 on Build GRCh38
rs887327382 Chr.21: 14593993-14593993 on Build GRCh38
rs766258909 Chr.21: 14593974-14593974 on Build GRCh38
rs778935157 Chr.21: 14594065-14594065 on Build GRCh38
rs576497710 Chr.21: 14594029-14594029 on Build GRCh38
rs993185564 Chr.21: 14593954-14593954 on Build GRCh38
rs769073434 Chr.21: 14594120-14594120 on Build GRCh38

View Full Product Details