Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 75584683-75584908 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLK
Entrez Gene ID
51426
Gene Name
polymerase (DNA) kappa
Gene Aliases
DINB1, DINP, POLQ
UniGene
Hs.135756
Gene Chromosome Location
Chr.5: 75511832-75602576 on Build GRCh38
Transcripts
NM_016218.2 XM_005248534.4 XM_011543463.2 XM_011543464.2 XM_011543467.2 XM_017009558.1 XM_017009559.1 XM_017009560.1 XM_017009561.1 XM_017009562.1 XM_017009563.1 XM_017009564.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6104308 c.1197C>T p.S399S Chr.5: 75584897-75584897 on Build GRCh38
5604557 c.1100C>T p.A367V Chr.5: 75584800-75584800 on Build GRCh38
7384916 c.1132C>T p.Q378* Chr.5: 75584832-75584832 on Build GRCh38
1070125 c.1147C>T p.L383F Chr.5: 75584847-75584847 on Build GRCh38
3776841 c.1112T>C p.I371T Chr.5: 75584812-75584812 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764444292 Chr.5: 75584832-75584832 on Build GRCh38
rs547011939 Chr.5: 75584863-75584863 on Build GRCh38
rs149529551 Chr.5: 75584844-75584844 on Build GRCh38
rs751790040 Chr.5: 75584730-75584732 on Build GRCh38
rs766166014 Chr.5: 75584862-75584862 on Build GRCh38
rs757994841 Chr.5: 75584875-75584875 on Build GRCh38
rs770269433 Chr.5: 75584811-75584811 on Build GRCh38
rs138077589 Chr.5: 75584719-75584719 on Build GRCh38
rs200557500 Chr.5: 75584774-75584774 on Build GRCh38
rs891644538 Chr.5: 75584738-75584738 on Build GRCh38
rs777198545 Chr.5: 75584799-75584799 on Build GRCh38
rs751546391 Chr.5: 75584834-75584834 on Build GRCh38
rs772666412 Chr.5: 75584788-75584788 on Build GRCh38
rs1035353121 Chr.5: 75584840-75584840 on Build GRCh38
rs771283940 Chr.5: 75584795-75584795 on Build GRCh38
rs960097666 Chr.5: 75584842-75584842 on Build GRCh38
rs5744683 Chr.5: 75584783-75584783 on Build GRCh38
rs1051540022 Chr.5: 75584831-75584831 on Build GRCh38
rs755966865 Chr.5: 75584720-75584720 on Build GRCh38
rs759820400 Chr.5: 75584754-75584758 on Build GRCh38
rs752517000 Chr.5: 75584871-75584871 on Build GRCh38
rs776047465 Chr.5: 75584818-75584818 on Build GRCh38
rs754809579 Chr.5: 75584757-75584757 on Build GRCh38
rs771749538 Chr.5: 75584786-75584786 on Build GRCh38
rs1021573610 Chr.5: 75584751-75584751 on Build GRCh38
rs182404921 Chr.5: 75584766-75584766 on Build GRCh38
rs1011387585 Chr.5: 75584747-75584747 on Build GRCh38
rs761970289 Chr.5: 75584848-75584848 on Build GRCh38
rs760027871 Chr.5: 75584804-75584804 on Build GRCh38
rs763237470 Chr.5: 75584823-75584823 on Build GRCh38
rs141290112 Chr.5: 75584870-75584870 on Build GRCh38
rs911603025 Chr.5: 75584847-75584847 on Build GRCh38
rs931611935 Chr.5: 75584798-75584798 on Build GRCh38
rs746549263 Chr.5: 75584791-75584791 on Build GRCh38
rs780064747 Chr.5: 75584721-75584721 on Build GRCh38
rs748980614 Chr.5: 75584745-75584745 on Build GRCh38

View Full Product Details