Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 15049688-15049913 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OLAH
Entrez Gene ID
55301
Gene Name
oleoyl-ACP hydrolase
Gene Aliases
AURA1, SAST, THEDC1
UniGene
Hs.24309
Gene Chromosome Location
Chr.10: 15043896-15073852 on Build GRCh38
Transcripts
NM_001039702.2 NM_018324.2 XM_017016376.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1636901 c.101G>A p.W34* Chr.10: 15049703-15049703 on Build GRCh38
916646 c.163G>T p.E55* Chr.10: 15049765-15049765 on Build GRCh38
7380993 c.116C>A p.S39Y Chr.10: 15049718-15049718 on Build GRCh38
6247386 c.197G>T p.W66L Chr.10: 15049736-15049736 on Build GRCh38
6247385 c.134G>T p.W45L Chr.10: 15049736-15049736 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770758093 Chr.10: 15049745-15049745 on Build GRCh38
rs990274294 Chr.10: 15049709-15049709 on Build GRCh38
rs901168179 Chr.10: 15049710-15049710 on Build GRCh38
rs150151605 Chr.10: 15049783-15049783 on Build GRCh38
rs746825688 Chr.10: 15049744-15049744 on Build GRCh38
rs773663673 Chr.10: 15049813-15049813 on Build GRCh38
rs1014916972 Chr.10: 15049851-15049851 on Build GRCh38
rs372961669 Chr.10: 15049806-15049806 on Build GRCh38
rs754746056 Chr.10: 15049880-15049880 on Build GRCh38
rs777502655 Chr.10: 15049736-15049736 on Build GRCh38
rs749043533 Chr.10: 15049853-15049853 on Build GRCh38
rs745573485 Chr.10: 15049750-15049750 on Build GRCh38
rs573204198 Chr.10: 15049730-15049730 on Build GRCh38
rs755059554 Chr.10: 15049725-15049725 on Build GRCh38
rs1024545924 Chr.10: 15049867-15049867 on Build GRCh38
rs981659070 Chr.10: 15049761-15049761 on Build GRCh38
rs769333505 Chr.10: 15049762-15049762 on Build GRCh38
rs7909465 Chr.10: 15049837-15049837 on Build GRCh38
rs371826903 Chr.10: 15049746-15049746 on Build GRCh38
rs1048832322 Chr.10: 15049847-15049847 on Build GRCh38
rs897548509 Chr.10: 15049820-15049820 on Build GRCh38
rs894644634 Chr.10: 15049839-15049839 on Build GRCh38
rs368669175 Chr.10: 15049734-15049734 on Build GRCh38
rs561777124 Chr.10: 15049819-15049819 on Build GRCh38
rs774873536 Chr.10: 15049763-15049763 on Build GRCh38
rs560224268 Chr.10: 15049765-15049765 on Build GRCh38
rs888509932 Chr.10: 15049787-15049787 on Build GRCh38

View Full Product Details