Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 7873474-7873737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTRR
Entrez Gene ID
4552
Gene Name
5-methyltetrahydrofolate-homocysteine methyltransferase reductase
Gene Aliases
MSR, cblE
UniGene
Hs.481551
Gene Chromosome Location
Chr.5: 7869104-7901124 on Build GRCh38
Transcripts
NM_002454.2 NM_024010.2 NR_134480.1 NR_134481.1 NR_134482.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6716969 c.358T>C p.L120L Chr.5: 7873520-7873520 on Build GRCh38
5756702 c.316C>G p.Q106E Chr.5: 7873478-7873478 on Build GRCh38
1695366 c.364G>A p.G122S Chr.5: 7873526-7873526 on Build GRCh38
5466565 c.336C>A p.F112L Chr.5: 7873498-7873498 on Build GRCh38
6171932 c.326C>T p.P109L Chr.5: 7873488-7873488 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143579470 Chr.5: 7873502-7873502 on Build GRCh38
rs781339117 Chr.5: 7873568-7873568 on Build GRCh38
rs987194135 Chr.5: 7873701-7873701 on Build GRCh38
rs770540418 Chr.5: 7873507-7873507 on Build GRCh38
rs754287494 Chr.5: 7873536-7873536 on Build GRCh38
rs755870497 Chr.5: 7873571-7873571 on Build GRCh38
rs375951634 Chr.5: 7873547-7873547 on Build GRCh38
rs769637305 Chr.5: 7873498-7873498 on Build GRCh38
rs374726994 Chr.5: 7873594-7873595 on Build GRCh38
rs561816162 Chr.5: 7873652-7873652 on Build GRCh38
rs759311230 Chr.5: 7873512-7873512 on Build GRCh38
rs747906783 Chr.5: 7873569-7873569 on Build GRCh38
rs983819264 Chr.5: 7873635-7873635 on Build GRCh38
rs911630607 Chr.5: 7873713-7873713 on Build GRCh38
rs989381279 Chr.5: 7873632-7873632 on Build GRCh38
rs958188637 Chr.5: 7873581-7873581 on Build GRCh38
rs908475089 Chr.5: 7873658-7873658 on Build GRCh38
rs766507273 Chr.5: 7873553-7873553 on Build GRCh38
rs1030804278 Chr.5: 7873603-7873603 on Build GRCh38
rs368717215 Chr.5: 7873541-7873541 on Build GRCh38
rs765813808 Chr.5: 7873660-7873660 on Build GRCh38
rs138167806 Chr.5: 7873504-7873504 on Build GRCh38
rs755219927 Chr.5: 7873562-7873562 on Build GRCh38
rs767247740 Chr.5: 7873515-7873515 on Build GRCh38
rs970744542 Chr.5: 7873531-7873531 on Build GRCh38
rs368391537 Chr.5: 7873539-7873539 on Build GRCh38
rs750878635 Chr.5: 7873549-7873549 on Build GRCh38
rs149323908 Chr.5: 7873555-7873555 on Build GRCh38
rs953215544 Chr.5: 7873653-7873653 on Build GRCh38
rs939932363 Chr.5: 7873682-7873682 on Build GRCh38
rs1025032500 Chr.5: 7873524-7873524 on Build GRCh38
rs78429440 Chr.5: 7873636-7873636 on Build GRCh38
rs773984668 Chr.5: 7873511-7873511 on Build GRCh38
rs548813608 Chr.5: 7873626-7873626 on Build GRCh38

View Full Product Details