Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1179959-1180222 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTLL10
Entrez Gene ID
254173
Gene Name
tubulin tyrosine ligase like 10
Gene Aliases
TTLL5
UniGene
Hs.454835
Gene Chromosome Location
Chr.1: 1173709-1197936 on Build GRCh38
Transcripts
NM_001130045.1 NM_153254.2 XM_005244738.1 XM_011541177.2 XM_017000906.1 XM_017000907.1 XM_017000908.1 XM_017000909.1 XM_017000910.1 XM_017000911.1 XM_017000912.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4973956 c.28C>T p.R10W Chr.1: 1180081-1180081 on Build GRCh38
6459115 c.387C>T p.N129N Chr.1: 1180221-1180221 on Build GRCh38
6409043 c.55G>T p.D19Y Chr.1: 1180108-1180108 on Build GRCh38
1686865 c.307G>A p.G103R Chr.1: 1180141-1180141 on Build GRCh38
6432047 c.349C>A p.L117M Chr.1: 1180183-1180183 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11811498 Chr.1: 1180074-1180074 on Build GRCh38
rs756766533 Chr.1: 1180050-1180050 on Build GRCh38
rs201647048 Chr.1: 1180194-1180194 on Build GRCh38
rs568644278 Chr.1: 1179998-1179998 on Build GRCh38
rs748355522 Chr.1: 1180136-1180136 on Build GRCh38
rs373208551 Chr.1: 1180132-1180132 on Build GRCh38
rs144432977 Chr.1: 1180182-1180182 on Build GRCh38
rs939918194 Chr.1: 1180054-1180054 on Build GRCh38
rs768804708 Chr.1: 1180022-1180022 on Build GRCh38
rs116383664 Chr.1: 1180081-1180081 on Build GRCh38
rs774183892 Chr.1: 1180195-1180195 on Build GRCh38
rs772262149 Chr.1: 1180082-1180082 on Build GRCh38
rs772461563 Chr.1: 1180143-1180143 on Build GRCh38
rs111649714 Chr.1: 1179999-1179999 on Build GRCh38
rs774436803 Chr.1: 1180075-1180075 on Build GRCh38
rs539820878 Chr.1: 1180141-1180141 on Build GRCh38
rs967705931 Chr.1: 1180021-1180021 on Build GRCh38
rs767192114 Chr.1: 1180198-1180198 on Build GRCh38
rs199586129 Chr.1: 1180175-1180175 on Build GRCh38
rs568320285 Chr.1: 1180164-1180165 on Build GRCh38
rs267597918 Chr.1: 1180173-1180173 on Build GRCh38
rs956409176 Chr.1: 1180121-1180121 on Build GRCh38
rs148494067 Chr.1: 1180016-1180016 on Build GRCh38
rs916160576 Chr.1: 1179979-1179979 on Build GRCh38
rs142661836 Chr.1: 1180196-1180196 on Build GRCh38
rs745864275 Chr.1: 1180171-1180171 on Build GRCh38
rs770337140 Chr.1: 1180067-1180067 on Build GRCh38
rs558272914 Chr.1: 1180045-1180045 on Build GRCh38
rs114390380 Chr.1: 1180168-1180168 on Build GRCh38
rs376545249 Chr.1: 1180038-1180038 on Build GRCh38
rs765057142 Chr.1: 1180036-1180036 on Build GRCh38
rs773465793 Chr.1: 1180031-1180031 on Build GRCh38
rs373544277 Chr.1: 1180018-1180018 on Build GRCh38
rs749649669 Chr.1: 1180072-1180072 on Build GRCh38
rs766995610 Chr.1: 1180122-1180122 on Build GRCh38
rs111751804 Chr.1: 1180123-1180123 on Build GRCh38
rs746363680 Chr.1: 1180058-1180058 on Build GRCh38
rs765117927 Chr.1: 1180088-1180088 on Build GRCh38
rs746167105 Chr.1: 1180009-1180009 on Build GRCh38
rs369075338 Chr.1: 1180026-1180026 on Build GRCh38
rs775306451 Chr.1: 1180107-1180107 on Build GRCh38
rs147884723 Chr.1: 1180084-1180084 on Build GRCh38
rs769667420 Chr.1: 1180189-1180189 on Build GRCh38
rs751211717 Chr.1: 1180111-1180111 on Build GRCh38
rs752475115 Chr.1: 1180037-1180037 on Build GRCh38
rs937781227 Chr.1: 1180029-1180029 on Build GRCh38
rs756608588 Chr.1: 1180128-1180128 on Build GRCh38
rs780439581 Chr.1: 1180069-1180069 on Build GRCh38
rs983793806 Chr.1: 1180188-1180188 on Build GRCh38
rs756962851 Chr.1: 1180118-1180118 on Build GRCh38
rs57556493 Chr.1: 1180035-1180035 on Build GRCh38
rs780748749 Chr.1: 1180056-1180056 on Build GRCh38
rs200681528 Chr.1: 1180025-1180025 on Build GRCh38
rs763735733 Chr.1: 1180110-1180110 on Build GRCh38
rs779476640 Chr.1: 1180135-1180135 on Build GRCh38
rs773383865 Chr.1: 1180167-1180167 on Build GRCh38
rs777313895 Chr.1: 1180044-1180044 on Build GRCh38
rs781097397 Chr.1: 1180168-1180168 on Build GRCh38
rs956288725 Chr.1: 1180010-1180010 on Build GRCh38
rs759513559 Chr.1: 1180086-1180086 on Build GRCh38
rs1000132549 Chr.1: 1179978-1179978 on Build GRCh38
rs114359609 Chr.1: 1180130-1180130 on Build GRCh38
rs770140865 Chr.1: 1180013-1180013 on Build GRCh38
rs377618504 Chr.1: 1180155-1180155 on Build GRCh38
rs370591688 Chr.1: 1180108-1180108 on Build GRCh38

View Full Product Details