Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 26365122-26365357 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF683
Entrez Gene ID
257101
Gene Name
zinc finger protein 683
Gene Aliases
Hobit
UniGene
Hs.353208
Gene Chromosome Location
Chr.1: 26361632-26374520 on Build GRCh38
Transcripts
NM_001114759.2 NM_001307925.1 NM_173574.3 XM_005245828.3 XM_005245830.3 XM_005245832.3 XM_006710555.3 XM_011541198.2 XM_017000954.1 XM_017000955.1 XM_017000956.1 XM_017000957.1 XM_017000958.1 XM_017000959.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2235253 c.352A>G p.T118A Chr.1: 26365194-26365194 on Build GRCh38
6312628 c.324C>A p.H108Q Chr.1: 26365222-26365222 on Build GRCh38
907662 c.276G>A p.K92K Chr.1: 26365225-26365225 on Build GRCh38
1341352 c.310G>A p.D104N Chr.1: 26365191-26365191 on Build GRCh38
3966490 c.316A>G p.K106E Chr.1: 26365185-26365185 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs534149045 Chr.1: 26365207-26365207 on Build GRCh38
rs780898539 Chr.1: 26365218-26365218 on Build GRCh38
rs886844546 Chr.1: 26365215-26365215 on Build GRCh38
rs748915921 Chr.1: 26365232-26365232 on Build GRCh38
rs752747857 Chr.1: 26365188-26365188 on Build GRCh38
rs758910989 Chr.1: 26365282-26365282 on Build GRCh38
rs200107113 Chr.1: 26365154-26365154 on Build GRCh38
rs1029423241 Chr.1: 26365308-26365308 on Build GRCh38
rs574772802 Chr.1: 26365202-26365202 on Build GRCh38
rs372687717 Chr.1: 26365173-26365173 on Build GRCh38
rs147764170 Chr.1: 26365214-26365214 on Build GRCh38
rs142231514 Chr.1: 26365192-26365192 on Build GRCh38
rs143491364 Chr.1: 26365222-26365222 on Build GRCh38
rs766702377 Chr.1: 26365176-26365176 on Build GRCh38
rs187778427 Chr.1: 26365159-26365159 on Build GRCh38
rs370102430 Chr.1: 26365247-26365247 on Build GRCh38
rs545133649 Chr.1: 26365303-26365303 on Build GRCh38
rs149941490 Chr.1: 26365174-26365174 on Build GRCh38
rs572851961 Chr.1: 26365283-26365283 on Build GRCh38
rs758139138 Chr.1: 26365199-26365199 on Build GRCh38
rs760028437 Chr.1: 26365185-26365185 on Build GRCh38
rs890441105 Chr.1: 26365319-26365319 on Build GRCh38
rs746883873 Chr.1: 26365144-26365144 on Build GRCh38
rs747684398 Chr.1: 26365221-26365221 on Build GRCh38
rs1055875352 Chr.1: 26365161-26365161 on Build GRCh38
rs74061129 Chr.1: 26365146-26365146 on Build GRCh38
rs1038162493 Chr.1: 26365243-26365243 on Build GRCh38
rs774855833 Chr.1: 26365179-26365179 on Build GRCh38
rs776241295 Chr.1: 26365246-26365247 on Build GRCh38
rs192637423 Chr.1: 26365246-26365246 on Build GRCh38
rs373823990 Chr.1: 26365191-26365191 on Build GRCh38
rs951538899 Chr.1: 26365329-26365329 on Build GRCh38
rs761597760 Chr.1: 26365156-26365156 on Build GRCh38
rs772748520 Chr.1: 26365225-26365225 on Build GRCh38

View Full Product Details