Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 74738737-74738994 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TYW3
Entrez Gene ID
127253
Gene Name
tRNA-yW synthesizing protein 3 homolog
Gene Aliases
C1orf171
UniGene
Hs.348411
Gene Chromosome Location
Chr.1: 74733143-74766677 on Build GRCh38
Transcripts
NM_001162916.1 NM_138467.2 NR_027962.1 XM_006710347.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4738556 c.337C>T p.Q113* Chr.1: 74738771-74738771 on Build GRCh38
1344295 c.304C>T p.P102S Chr.1: 74738738-74738738 on Build GRCh38
78736 c.305C>G p.P102R Chr.1: 74738739-74738739 on Build GRCh38
682121 c.305C>T p.P102L Chr.1: 74738739-74738739 on Build GRCh38
6064359 c.336G>A p.L112L Chr.1: 74738770-74738770 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs527814467 Chr.1: 74738872-74738872 on Build GRCh38
rs1031389124 Chr.1: 74738956-74738956 on Build GRCh38
rs760957824 Chr.1: 74738825-74738825 on Build GRCh38
rs570800824 Chr.1: 74738880-74738880 on Build GRCh38
rs944513944 Chr.1: 74738901-74738901 on Build GRCh38
rs1042222735 Chr.1: 74738883-74738883 on Build GRCh38
rs773250951 Chr.1: 74738824-74738824 on Build GRCh38
rs768387030 Chr.1: 74738896-74738896 on Build GRCh38
rs552615965 Chr.1: 74738877-74738877 on Build GRCh38
rs888912818 Chr.1: 74738899-74738899 on Build GRCh38
rs374951745 Chr.1: 74738920-74738920 on Build GRCh38
rs556904239 Chr.1: 74738925-74738925 on Build GRCh38
rs749751137 Chr.1: 74738794-74738794 on Build GRCh38
rs920536743 Chr.1: 74738857-74738857 on Build GRCh38
rs11538281 Chr.1: 74738763-74738763 on Build GRCh38
rs755560668 Chr.1: 74738762-74738762 on Build GRCh38
rs772549022 Chr.1: 74738814-74738814 on Build GRCh38
rs766527082 Chr.1: 74738832-74738832 on Build GRCh38
rs566969475 Chr.1: 74738853-74738853 on Build GRCh38
rs940385615 Chr.1: 74738801-74738801 on Build GRCh38
rs748459266 Chr.1: 74738812-74738812 on Build GRCh38
rs774966739 Chr.1: 74738808-74738808 on Build GRCh38
rs376274015 Chr.1: 74738775-74738775 on Build GRCh38
rs3736407 Chr.1: 74738841-74738841 on Build GRCh38
rs769132790 Chr.1: 74738799-74738799 on Build GRCh38
rs971692886 Chr.1: 74738855-74738855 on Build GRCh38
rs927717809 Chr.1: 74738766-74738766 on Build GRCh38
rs1054144374 Chr.1: 74738787-74738787 on Build GRCh38
rs1041189161 Chr.1: 74738910-74738910 on Build GRCh38
rs945233716 Chr.1: 74738876-74738876 on Build GRCh38

View Full Product Details