Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 156954322-156954593 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TIMD4
Entrez Gene ID
91937
Gene Name
T-cell immunoglobulin and mucin domain containing 4
Gene Aliases
SMUCKLER, TIM4
UniGene
Hs.334907
Gene Chromosome Location
Chr.5: 156915750-156963255 on Build GRCh38
Transcripts
NM_001146726.1 NM_138379.2 XM_011534694.2 XM_017010021.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7106808 c.236G>T p.R79I Chr.5: 156954579-156954579 on Build GRCh38
1310891 c.273G>A p.P91P Chr.5: 156954542-156954542 on Build GRCh38
4902964 c.391C>T p.L131L Chr.5: 156954424-156954424 on Build GRCh38
6102655 c.234A>T p.S78S Chr.5: 156954581-156954581 on Build GRCh38
3919453 c.364G>A p.D122N Chr.5: 156954451-156954451 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs187461354 Chr.5: 156954386-156954386 on Build GRCh38
rs756535305 Chr.5: 156954408-156954408 on Build GRCh38
rs766204987 Chr.5: 156954452-156954452 on Build GRCh38
rs751158268 Chr.5: 156954389-156954389 on Build GRCh38
rs112267968 Chr.5: 156954440-156954440 on Build GRCh38
rs754265826 Chr.5: 156954471-156954471 on Build GRCh38
rs749628221 Chr.5: 156954427-156954427 on Build GRCh38
rs761841993 Chr.5: 156954437-156954437 on Build GRCh38
rs367759946 Chr.5: 156954494-156954494 on Build GRCh38
rs754624689 Chr.5: 156954455-156954455 on Build GRCh38
rs762814005 Chr.5: 156954451-156954451 on Build GRCh38
rs1018069747 Chr.5: 156954358-156954358 on Build GRCh38
rs778218447 Chr.5: 156954414-156954414 on Build GRCh38
rs139335693 Chr.5: 156954477-156954477 on Build GRCh38
rs775366250 Chr.5: 156954436-156954436 on Build GRCh38
rs377691244 Chr.5: 156954456-156954456 on Build GRCh38
rs926482965 Chr.5: 156954354-156954354 on Build GRCh38
rs752096886 Chr.5: 156954462-156954462 on Build GRCh38
rs761597210 Chr.5: 156954372-156954372 on Build GRCh38
rs933292060 Chr.5: 156954545-156954545 on Build GRCh38
rs770849831 Chr.5: 156954539-156954539 on Build GRCh38
rs751914510 Chr.5: 156954390-156954390 on Build GRCh38
rs780206539 Chr.5: 156954496-156954496 on Build GRCh38
rs533342571 Chr.5: 156954558-156954558 on Build GRCh38
rs758148694 Chr.5: 156954387-156954387 on Build GRCh38
rs556271265 Chr.5: 156954362-156954362 on Build GRCh38
rs773192900 Chr.5: 156954449-156954449 on Build GRCh38
rs774244018 Chr.5: 156954542-156954542 on Build GRCh38
rs778131170 Chr.5: 156954347-156954347 on Build GRCh38
rs7724832 Chr.5: 156954359-156954359 on Build GRCh38
rs760092945 Chr.5: 156954563-156954563 on Build GRCh38
rs748689172 Chr.5: 156954405-156954405 on Build GRCh38
rs116394296 Chr.5: 156954543-156954543 on Build GRCh38
rs371298031 Chr.5: 156954464-156954464 on Build GRCh38
rs917689907 Chr.5: 156954367-156954367 on Build GRCh38
rs746095229 Chr.5: 156954478-156954478 on Build GRCh38
rs751248021 Chr.5: 156954453-156954453 on Build GRCh38
rs771181155 Chr.5: 156954431-156954431 on Build GRCh38
rs778109353 Chr.5: 156954466-156954466 on Build GRCh38
rs1019048919 Chr.5: 156954534-156954534 on Build GRCh38
rs141557472 Chr.5: 156954433-156954433 on Build GRCh38
rs773281432 Chr.5: 156954537-156954537 on Build GRCh38
rs747038780 Chr.5: 156954509-156954509 on Build GRCh38
rs754534905 Chr.5: 156954393-156954393 on Build GRCh38
rs774551130 Chr.5: 156954432-156954432 on Build GRCh38
rs146762720 Chr.5: 156954538-156954538 on Build GRCh38
rs867149071 Chr.5: 156954567-156954567 on Build GRCh38
rs767048599 Chr.5: 156954546-156954546 on Build GRCh38
rs991857388 Chr.5: 156954472-156954472 on Build GRCh38
rs776616868 Chr.5: 156954370-156954370 on Build GRCh38
rs769812788 Chr.5: 156954522-156954522 on Build GRCh38
rs373602957 Chr.5: 156954479-156954479 on Build GRCh38
rs889310322 Chr.5: 156954491-156954491 on Build GRCh38
rs781611375 Chr.5: 156954400-156954400 on Build GRCh38
rs764654169 Chr.5: 156954568-156954568 on Build GRCh38
rs148723473 Chr.5: 156954417-156954417 on Build GRCh38
rs767082333 Chr.5: 156954460-156954460 on Build GRCh38

View Full Product Details