Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 50128122-50128381 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM189
Entrez Gene ID
387521
Gene Name
transmembrane protein 189
Gene Aliases
KUA
UniGene
Hs.744839
Gene Chromosome Location
Chr.20: 50123737-50153798 on Build GRCh38
Transcripts
NM_001162505.1 NM_199129.2 NR_027889.1 NM_199203.2

Gene Symbol
TMEM189-UBE2V1
Entrez Gene ID
387522
Gene Name
TMEM189-UBE2V1 readthrough
Gene Aliases
CROC-1B, CROC1B, KUA-UEV
UniGene
Hs.744839
Gene Chromosome Location
Chr.20: 50081124-50153798 on Build GRCh38
Transcripts
NM_001162505.1 NM_199129.2 NR_027889.1 NM_199203.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5366017 c.528C>A p.I176I Chr.20: 50128138-50128138 on Build GRCh38
5366016 c.528C>A p.I176I Chr.20: 50128138-50128138 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs543087302 Chr.20: 50128317-50128317 on Build GRCh38
rs141662240 Chr.20: 50128212-50128212 on Build GRCh38
rs554489753 Chr.20: 50128213-50128213 on Build GRCh38
rs747191244 Chr.20: 50128217-50128217 on Build GRCh38
rs771166583 Chr.20: 50128168-50128168 on Build GRCh38
rs567143775 Chr.20: 50128203-50128203 on Build GRCh38
rs774524205 Chr.20: 50128177-50128177 on Build GRCh38
rs749603950 Chr.20: 50128288-50128288 on Build GRCh38
rs368258608 Chr.20: 50128233-50128233 on Build GRCh38
rs749753581 Chr.20: 50128159-50128159 on Build GRCh38
rs528845990 Chr.20: 50128342-50128342 on Build GRCh38
rs576094834 Chr.20: 50128314-50128314 on Build GRCh38
rs775591336 Chr.20: 50128215-50128215 on Build GRCh38
rs895235855 Chr.20: 50128312-50128312 on Build GRCh38
rs758954026 Chr.20: 50128239-50128239 on Build GRCh38
rs370448658 Chr.20: 50128204-50128204 on Build GRCh38
rs772938906 Chr.20: 50128228-50128228 on Build GRCh38
rs143619190 Chr.20: 50128184-50128184 on Build GRCh38
rs572577107 Chr.20: 50128276-50128276 on Build GRCh38
rs945994725 Chr.20: 50128260-50128260 on Build GRCh38
rs6091099 Chr.20: 50128294-50128294 on Build GRCh38
rs991665687 Chr.20: 50128241-50128241 on Build GRCh38
rs931059117 Chr.20: 50128192-50128192 on Build GRCh38
rs756447665 Chr.20: 50128209-50128209 on Build GRCh38
rs753162354 Chr.20: 50128207-50128207 on Build GRCh38
rs899080810 Chr.20: 50128336-50128336 on Build GRCh38
rs376087874 Chr.20: 50128194-50128194 on Build GRCh38
rs769612057 Chr.20: 50128226-50128226 on Build GRCh38
rs761503001 Chr.20: 50128200-50128200 on Build GRCh38
rs773975556 Chr.20: 50128236-50128236 on Build GRCh38
rs766898063 Chr.20: 50128275-50128275 on Build GRCh38
rs770253572 Chr.20: 50128278-50128278 on Build GRCh38
rs772398533 Chr.20: 50128214-50128214 on Build GRCh38
rs62209907 Chr.20: 50128325-50128325 on Build GRCh38
rs200277814 Chr.20: 50128143-50128143 on Build GRCh38
rs756709177 Chr.20: 50128152-50128152 on Build GRCh38
rs764958624 Chr.20: 50128202-50128202 on Build GRCh38
rs148716126 Chr.20: 50128150-50128150 on Build GRCh38
rs926109399 Chr.20: 50128234-50128234 on Build GRCh38
rs527913597 Chr.20: 50128203-50128204 on Build GRCh38
rs758999773 Chr.20: 50128204-50128204 on Build GRCh38
rs746152559 Chr.20: 50128179-50128179 on Build GRCh38
rs766269630 Chr.20: 50128234-50128234 on Build GRCh38
rs773143306 Chr.20: 50128203-50128203 on Build GRCh38
rs6095774 Chr.20: 50128281-50128281 on Build GRCh38
rs138028944 Chr.20: 50128205-50128205 on Build GRCh38
rs887144532 Chr.20: 50128301-50128301 on Build GRCh38
rs374299929 Chr.20: 50128210-50128210 on Build GRCh38
rs778410937 Chr.20: 50128153-50128153 on Build GRCh38

View Full Product Details