Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 53254207-53254438 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSHZ2
Entrez Gene ID
128553
Gene Name
teashirt zinc finger homeobox 2
Gene Aliases
C20orf17, OVC10-2, TSH2, ZABC2, ZNF218
UniGene
Hs.473117
Gene Chromosome Location
Chr.20: 52972309-53495330 on Build GRCh38
Transcripts
NM_001193421.1 NM_173485.5 XM_017027640.1 XM_017027641.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1580018 c.882G>A p.M294I Chr.20: 53254340-53254340 on Build GRCh38
258976 c.844G>A p.D282N Chr.20: 53254302-53254302 on Build GRCh38
5540968 c.789C>T p.F263F Chr.20: 53254247-53254247 on Build GRCh38
7395441 c.773C>A p.P258H Chr.20: 53254231-53254231 on Build GRCh38
6525952 c.911C>A p.P304H Chr.20: 53254369-53254369 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751286817 Chr.20: 53254278-53254278 on Build GRCh38
rs749729408 Chr.20: 53254414-53254414 on Build GRCh38
rs752188288 Chr.20: 53254400-53254400 on Build GRCh38
rs750169163 Chr.20: 53254254-53254254 on Build GRCh38
rs751446698 Chr.20: 53254392-53254392 on Build GRCh38
rs920721673 Chr.20: 53254368-53254368 on Build GRCh38
rs771232902 Chr.20: 53254327-53254327 on Build GRCh38
rs373677633 Chr.20: 53254376-53254376 on Build GRCh38
rs766055253 Chr.20: 53254383-53254383 on Build GRCh38
rs774739292 Chr.20: 53254330-53254330 on Build GRCh38
rs564692924 Chr.20: 53254304-53254304 on Build GRCh38
rs754663526 Chr.20: 53254397-53254397 on Build GRCh38
rs765161050 Chr.20: 53254379-53254379 on Build GRCh38
rs755602779 Chr.20: 53254412-53254412 on Build GRCh38
rs772340421 Chr.20: 53254332-53254332 on Build GRCh38
rs767343963 Chr.20: 53254399-53254399 on Build GRCh38
rs141028930 Chr.20: 53254326-53254326 on Build GRCh38
rs573494662 Chr.20: 53254331-53254331 on Build GRCh38
rs739870 Chr.20: 53254310-53254310 on Build GRCh38
rs763187600 Chr.20: 53254276-53254276 on Build GRCh38
rs373894649 Chr.20: 53254244-53254244 on Build GRCh38
rs780826702 Chr.20: 53254302-53254302 on Build GRCh38
rs762766937 Chr.20: 53254382-53254382 on Build GRCh38
rs765158720 Chr.20: 53254245-53254245 on Build GRCh38
rs754714953 Chr.20: 53254301-53254301 on Build GRCh38
rs758137267 Chr.20: 53254265-53254265 on Build GRCh38
rs142965743 Chr.20: 53254306-53254306 on Build GRCh38
rs773042192 Chr.20: 53254381-53254381 on Build GRCh38
rs778714438 Chr.20: 53254320-53254320 on Build GRCh38
rs775547288 Chr.20: 53254337-53254337 on Build GRCh38
rs45562541 Chr.20: 53254415-53254415 on Build GRCh38

View Full Product Details