Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 37636999-37637272 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRPC4
Entrez Gene ID
7223
Gene Name
transient receptor potential cation channel subfamily C member 4
Gene Aliases
HTRP-4, HTRP4, TRP4
UniGene
Hs.262960
Gene Chromosome Location
Chr.13: 37633872-37870374 on Build GRCh38
Transcripts
NM_001135955.1 NM_001135956.1 NM_001135957.1 NM_001135958.1 NM_003306.1 NM_016179.2 XM_011535206.1 XM_017020723.1 XM_017020724.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2072863 c.2641G>T p.A881S Chr.13: 37637211-37637211 on Build GRCh38
4047469 c.2618G>A p.R873H Chr.13: 37637219-37637219 on Build GRCh38
3468704 c.2775G>A p.K925K Chr.13: 37637062-37637062 on Build GRCh38
947207 c.2629G>T p.A877S Chr.13: 37637223-37637223 on Build GRCh38
108185 c.2595C>T p.F865F Chr.13: 37637242-37637242 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760141036 Chr.13: 37637077-37637077 on Build GRCh38
rs539868562 Chr.13: 37637155-37637155 on Build GRCh38
rs772536839 Chr.13: 37637081-37637081 on Build GRCh38
rs752969074 Chr.13: 37637226-37637228 on Build GRCh38
rs142683062 Chr.13: 37637219-37637219 on Build GRCh38
rs147399086 Chr.13: 37637169-37637169 on Build GRCh38
rs887382142 Chr.13: 37637084-37637084 on Build GRCh38
rs200985201 Chr.13: 37637122-37637122 on Build GRCh38
rs761827542 Chr.13: 37637106-37637106 on Build GRCh38
rs35395094 Chr.13: 37637152-37637152 on Build GRCh38
rs751551802 Chr.13: 37637059-37637059 on Build GRCh38
rs997127463 Chr.13: 37637185-37637185 on Build GRCh38
rs146885289 Chr.13: 37637242-37637242 on Build GRCh38
rs149495867 Chr.13: 37637062-37637062 on Build GRCh38
rs373682167 Chr.13: 37637029-37637029 on Build GRCh38
rs779347338 Chr.13: 37637068-37637068 on Build GRCh38
rs534937697 Chr.13: 37637241-37637241 on Build GRCh38
rs760658714 Chr.13: 37637186-37637186 on Build GRCh38
rs140515294 Chr.13: 37637096-37637096 on Build GRCh38
rs912360920 Chr.13: 37637208-37637208 on Build GRCh38
rs748682074 Chr.13: 37637239-37637239 on Build GRCh38
rs776121635 Chr.13: 37637090-37637090 on Build GRCh38
rs768211571 Chr.13: 37637252-37637252 on Build GRCh38
rs377352848 Chr.13: 37637111-37637111 on Build GRCh38
rs747573040 Chr.13: 37637082-37637082 on Build GRCh38
rs1048621556 Chr.13: 37637078-37637078 on Build GRCh38
rs769331202 Chr.13: 37637153-37637153 on Build GRCh38
rs778212234 Chr.13: 37637131-37637131 on Build GRCh38
rs114600063 Chr.13: 37637202-37637202 on Build GRCh38
rs767791117 Chr.13: 37637209-37637220 on Build GRCh38
rs369573111 Chr.13: 37637072-37637072 on Build GRCh38
rs375645285 Chr.13: 37637220-37637220 on Build GRCh38
rs958351197 Chr.13: 37637157-37637157 on Build GRCh38
rs751918556 Chr.13: 37637038-37637038 on Build GRCh38
rs771321099 Chr.13: 37637146-37637146 on Build GRCh38
rs75661457 Chr.13: 37637176-37637176 on Build GRCh38
rs555606878 Chr.13: 37637124-37637124 on Build GRCh38
rs531504926 Chr.13: 37637226-37637226 on Build GRCh38
rs559888954 Chr.13: 37637158-37637158 on Build GRCh38
rs757099137 Chr.13: 37637060-37637060 on Build GRCh38
rs370995069 Chr.13: 37637156-37637156 on Build GRCh38
rs751793234 Chr.13: 37637211-37637211 on Build GRCh38
rs2957221 Chr.13: 37637232-37637232 on Build GRCh38
rs758509700 Chr.13: 37637130-37637130 on Build GRCh38
rs759819042 Chr.13: 37637129-37637130 on Build GRCh38
rs775168860 Chr.13: 37637100-37637100 on Build GRCh38
rs770234760 Chr.13: 37637223-37637223 on Build GRCh38
rs771284130 Chr.13: 37637089-37637089 on Build GRCh38
rs758268382 Chr.13: 37637053-37637053 on Build GRCh38
rs753008221 Chr.13: 37637125-37637125 on Build GRCh38
rs377601240 Chr.13: 37637064-37637064 on Build GRCh38
rs763771971 Chr.13: 37637058-37637058 on Build GRCh38
rs35836067 Chr.13: 37637201-37637201 on Build GRCh38
rs781706314 Chr.13: 37637149-37637149 on Build GRCh38
rs373016207 Chr.13: 37637104-37637104 on Build GRCh38
rs765439306 Chr.13: 37637199-37637199 on Build GRCh38
rs147153854 Chr.13: 37637095-37637095 on Build GRCh38
rs1021535583 Chr.13: 37637177-37637177 on Build GRCh38
rs763977564 Chr.13: 37637102-37637102 on Build GRCh38
rs751755487 Chr.13: 37637047-37637047 on Build GRCh38
rs755167746 Chr.13: 37637117-37637117 on Build GRCh38
rs545536923 Chr.13: 37637196-37637196 on Build GRCh38
rs757032971 Chr.13: 37637241-37637243 on Build GRCh38
rs767498840 Chr.13: 37637116-37637116 on Build GRCh38
rs747269770 Chr.13: 37637135-37637135 on Build GRCh38
rs373986502 Chr.13: 37637172-37637172 on Build GRCh38
rs761582713 Chr.13: 37637171-37637171 on Build GRCh38

View Full Product Details