Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65116402-65116675 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNHIT2
Entrez Gene ID
741
Gene Name
zinc finger HIT-type containing 2
Gene Aliases
C11orf5, FON
UniGene
Hs.121025
Gene Chromosome Location
Chr.11: 65116403-65117738 on Build GRCh38
Transcripts
NM_014205.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5739310 c.988C>T p.R330* Chr.11: 65116666-65116666 on Build GRCh38
6833992 c.1167_1168insC p.V390fs*10 Chr.11: 65116486-65116487 on Build GRCh38
4035335 c.1113A>G p.V371V Chr.11: 65116541-65116541 on Build GRCh38
3791790 c.1078G>A p.D360N Chr.11: 65116576-65116576 on Build GRCh38
6833989 c.1016delA p.K339fs*21 Chr.11: 65116638-65116638 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779803219 Chr.11: 65116624-65116624 on Build GRCh38
rs750116186 Chr.11: 65116609-65116609 on Build GRCh38
rs1004875494 Chr.11: 65116433-65116433 on Build GRCh38
rs766970023 Chr.11: 65116507-65116507 on Build GRCh38
rs780825662 Chr.11: 65116537-65116537 on Build GRCh38
rs202078302 Chr.11: 65116574-65116574 on Build GRCh38
rs748734717 Chr.11: 65116564-65116564 on Build GRCh38
rs761171175 Chr.11: 65116581-65116581 on Build GRCh38
rs1546532 Chr.11: 65116580-65116580 on Build GRCh38
rs879273139 Chr.11: 65116495-65116495 on Build GRCh38
rs752873089 Chr.11: 65116598-65116598 on Build GRCh38
rs747771395 Chr.11: 65116571-65116571 on Build GRCh38
rs200656382 Chr.11: 65116486-65116486 on Build GRCh38
rs766708058 Chr.11: 65116549-65116549 on Build GRCh38
rs753859150 Chr.11: 65116517-65116517 on Build GRCh38
rs979684655 Chr.11: 65116531-65116531 on Build GRCh38
rs771277843 Chr.11: 65116582-65116582 on Build GRCh38
rs765376475 Chr.11: 65116595-65116595 on Build GRCh38
rs1037408275 Chr.11: 65116592-65116592 on Build GRCh38
rs777210354 Chr.11: 65116584-65116584 on Build GRCh38
rs376279736 Chr.11: 65116565-65116565 on Build GRCh38
rs764440850 Chr.11: 65116601-65116601 on Build GRCh38
rs755947643 Chr.11: 65116610-65116610 on Build GRCh38
rs771621208 Chr.11: 65116645-65116645 on Build GRCh38
rs767903789 Chr.11: 65116487-65116487 on Build GRCh38
rs149620690 Chr.11: 65116644-65116644 on Build GRCh38
rs747279780 Chr.11: 65116651-65116651 on Build GRCh38
rs1030106999 Chr.11: 65116646-65116646 on Build GRCh38
rs774846803 Chr.11: 65116446-65116446 on Build GRCh38
rs759640396 Chr.11: 65116587-65116587 on Build GRCh38
rs748893329 Chr.11: 65116480-65116480 on Build GRCh38
rs574434476 Chr.11: 65116501-65116501 on Build GRCh38
rs184318096 Chr.11: 65116504-65116504 on Build GRCh38
rs755999355 Chr.11: 65116546-65116546 on Build GRCh38
rs763863810 Chr.11: 65116440-65116440 on Build GRCh38
rs554127514 Chr.11: 65116431-65116431 on Build GRCh38
rs758732879 Chr.11: 65116534-65116534 on Build GRCh38
rs1018945110 Chr.11: 65116466-65116466 on Build GRCh38
rs777487612 Chr.11: 65116648-65116648 on Build GRCh38
rs373041278 Chr.11: 65116429-65116429 on Build GRCh38
rs374889298 Chr.11: 65116633-65116633 on Build GRCh38
rs752998089 Chr.11: 65116530-65116530 on Build GRCh38
rs745731608 Chr.11: 65116539-65116539 on Build GRCh38
rs755114348 Chr.11: 65116522-65116522 on Build GRCh38
rs771763995 Chr.11: 65116579-65116579 on Build GRCh38
rs144753523 Chr.11: 65116599-65116599 on Build GRCh38
rs754951975 Chr.11: 65116628-65116628 on Build GRCh38
rs370685853 Chr.11: 65116627-65116627 on Build GRCh38
rs556832705 Chr.11: 65116570-65116570 on Build GRCh38
rs780042211 Chr.11: 65116552-65116552 on Build GRCh38
rs745759821 Chr.11: 65116439-65116439 on Build GRCh38
rs781236042 Chr.11: 65116432-65116432 on Build GRCh38
rs768226850 Chr.11: 65116483-65116483 on Build GRCh38

View Full Product Details