Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 178520265-178520538 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TEX35
Entrez Gene ID
84066
Gene Name
testis expressed 35
Gene Aliases
C1orf49, TSC24
UniGene
Hs.534501
Gene Chromosome Location
Chr.1: 178513077-178523500 on Build GRCh38
Transcripts
NM_001170722.1 NM_001170723.1 NM_001170724.1 NM_032126.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5576467 c.301G>A p.E101K Chr.1: 178520372-178520372 on Build GRCh38
5576470 c.49G>A p.E17K Chr.1: 178520372-178520372 on Build GRCh38
5705052 c.104A>G p.N35S Chr.1: 178520427-178520427 on Build GRCh38
5705049 c.356A>G p.N119S Chr.1: 178520427-178520427 on Build GRCh38
5576469 c.277G>A p.E93K Chr.1: 178520372-178520372 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145745210 Chr.1: 178520416-178520416 on Build GRCh38
rs1042674942 Chr.1: 178520421-178520423 on Build GRCh38
rs753600663 Chr.1: 178520447-178520447 on Build GRCh38
rs770991713 Chr.1: 178520364-178520364 on Build GRCh38
rs766933498 Chr.1: 178520412-178520413 on Build GRCh38
rs759783335 Chr.1: 178520456-178520456 on Build GRCh38
rs376264869 Chr.1: 178520477-178520477 on Build GRCh38
rs775510387 Chr.1: 178520464-178520464 on Build GRCh38
rs370944731 Chr.1: 178520302-178520302 on Build GRCh38
rs565309035 Chr.1: 178520510-178520510 on Build GRCh38
rs143366830 Chr.1: 178520413-178520413 on Build GRCh38
rs763403408 Chr.1: 178520378-178520378 on Build GRCh38
rs578184505 Chr.1: 178520485-178520485 on Build GRCh38
rs753549981 Chr.1: 178520477-178520478 on Build GRCh38
rs769932240 Chr.1: 178520463-178520463 on Build GRCh38
rs773088882 Chr.1: 178520346-178520346 on Build GRCh38
rs376666368 Chr.1: 178520502-178520502 on Build GRCh38
rs759732968 Chr.1: 178520367-178520367 on Build GRCh38
rs201096376 Chr.1: 178520486-178520486 on Build GRCh38
rs751955974 Chr.1: 178520430-178520430 on Build GRCh38
rs200941431 Chr.1: 178520455-178520455 on Build GRCh38
rs757635093 Chr.1: 178520396-178520396 on Build GRCh38
rs778459640 Chr.1: 178520417-178520417 on Build GRCh38
rs746938788 Chr.1: 178520519-178520519 on Build GRCh38
rs139746729 Chr.1: 178520384-178520384 on Build GRCh38
rs146640002 Chr.1: 178520385-178520385 on Build GRCh38
rs757993536 Chr.1: 178520508-178520508 on Build GRCh38
rs758448375 Chr.1: 178520418-178520418 on Build GRCh38
rs373067711 Chr.1: 178520453-178520453 on Build GRCh38
rs375444894 Chr.1: 178520348-178520348 on Build GRCh38
rs767773730 Chr.1: 178520475-178520476 on Build GRCh38
rs746740772 Chr.1: 178520430-178520430 on Build GRCh38
rs754605229 Chr.1: 178520490-178520490 on Build GRCh38
rs201837136 Chr.1: 178520323-178520323 on Build GRCh38
rs370047377 Chr.1: 178520479-178520479 on Build GRCh38
rs765330762 Chr.1: 178520368-178520368 on Build GRCh38
rs764529754 Chr.1: 178520476-178520476 on Build GRCh38
rs376170105 Chr.1: 178520329-178520329 on Build GRCh38
rs763026707 Chr.1: 178520471-178520471 on Build GRCh38
rs759952897 Chr.1: 178520438-178520439 on Build GRCh38
rs777856044 Chr.1: 178520421-178520421 on Build GRCh38
rs200101991 Chr.1: 178520452-178520452 on Build GRCh38
rs917593383 Chr.1: 178520491-178520491 on Build GRCh38
rs770746739 Chr.1: 178520450-178520450 on Build GRCh38
rs765399942 Chr.1: 178520365-178520365 on Build GRCh38
rs575193524 Chr.1: 178520375-178520375 on Build GRCh38
rs929273377 Chr.1: 178520304-178520304 on Build GRCh38
rs781472263 Chr.1: 178520402-178520402 on Build GRCh38
rs752486799 Chr.1: 178520321-178520321 on Build GRCh38

View Full Product Details