Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 132570614-132570887 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAAR6
Entrez Gene ID
319100
Gene Name
trace amine associated receptor 6
Gene Aliases
TA4, TAR4, TAR6, TRAR4, taR-4, taR-6
UniGene
Hs.434196
Gene Chromosome Location
Chr.6: 132570322-132571359 on Build GRCh38
Transcripts
NM_175067.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3620725 c.449G>A p.G150E Chr.6: 132570770-132570770 on Build GRCh38
3620723 c.356C>T p.S119F Chr.6: 132570677-132570677 on Build GRCh38
6795874 c.400G>A p.V134I Chr.6: 132570721-132570721 on Build GRCh38
6172425 c.531G>T p.L177L Chr.6: 132570852-132570852 on Build GRCh38
4882645 c.478C>A p.P160T Chr.6: 132570799-132570799 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs71574624 Chr.6: 132570664-132570664 on Build GRCh38
rs781030582 Chr.6: 132570698-132570698 on Build GRCh38
rs371242104 Chr.6: 132570784-132570784 on Build GRCh38
rs909932467 Chr.6: 132570821-132570821 on Build GRCh38
rs777445168 Chr.6: 132570711-132570711 on Build GRCh38
rs745510373 Chr.6: 132570776-132570776 on Build GRCh38
rs150333502 Chr.6: 132570793-132570793 on Build GRCh38
rs768968103 Chr.6: 132570729-132570729 on Build GRCh38
rs201724538 Chr.6: 132570757-132570757 on Build GRCh38
rs754781633 Chr.6: 132570697-132570697 on Build GRCh38
rs776025242 Chr.6: 132570654-132570654 on Build GRCh38
rs766549957 Chr.6: 132570691-132570691 on Build GRCh38
rs184339469 Chr.6: 132570738-132570738 on Build GRCh38
rs201176105 Chr.6: 132570768-132570768 on Build GRCh38
rs778562117 Chr.6: 132570773-132570773 on Build GRCh38
rs758644231 Chr.6: 132570686-132570686 on Build GRCh38
rs773761203 Chr.6: 132570788-132570788 on Build GRCh38
rs770532856 Chr.6: 132570719-132570719 on Build GRCh38
rs748036227 Chr.6: 132570705-132570705 on Build GRCh38
rs530149491 Chr.6: 132570756-132570756 on Build GRCh38
rs759803881 Chr.6: 132570750-132570750 on Build GRCh38
rs565222153 Chr.6: 132570645-132570645 on Build GRCh38
rs762240830 Chr.6: 132570734-132570734 on Build GRCh38
rs760983898 Chr.6: 132570655-132570655 on Build GRCh38
rs764495674 Chr.6: 132570661-132570661 on Build GRCh38
rs969472791 Chr.6: 132570687-132570687 on Build GRCh38
rs958452124 Chr.6: 132570761-132570761 on Build GRCh38
rs770239115 Chr.6: 132570735-132570735 on Build GRCh38
rs757193971 Chr.6: 132570830-132570830 on Build GRCh38
rs1000808578 Chr.6: 132570746-132570746 on Build GRCh38
rs771069146 Chr.6: 132570790-132570790 on Build GRCh38
rs151162468 Chr.6: 132570663-132570663 on Build GRCh38
rs200377789 Chr.6: 132570813-132570813 on Build GRCh38
rs200184006 Chr.6: 132570751-132570751 on Build GRCh38
rs755041158 Chr.6: 132570706-132570706 on Build GRCh38
rs758020375 Chr.6: 132570841-132570841 on Build GRCh38
rs777062412 Chr.6: 132570662-132570662 on Build GRCh38
rs749325648 Chr.6: 132570649-132570649 on Build GRCh38
rs200733367 Chr.6: 132570732-132570732 on Build GRCh38
rs778756179 Chr.6: 132570720-132570720 on Build GRCh38
rs1026684369 Chr.6: 132570809-132570809 on Build GRCh38
rs763440114 Chr.6: 132570789-132570789 on Build GRCh38
rs942773699 Chr.6: 132570833-132570833 on Build GRCh38
rs376774841 Chr.6: 132570806-132570806 on Build GRCh38
rs751790102 Chr.6: 132570747-132570747 on Build GRCh38
rs770114428 Chr.6: 132570785-132570785 on Build GRCh38
rs530711693 Chr.6: 132570672-132570672 on Build GRCh38
rs776988020 Chr.6: 132570730-132570730 on Build GRCh38
rs745520897 Chr.6: 132570727-132570727 on Build GRCh38
rs751752246 Chr.6: 132570692-132570692 on Build GRCh38
rs140176422 Chr.6: 132570714-132570714 on Build GRCh38
rs368130988 Chr.6: 132570780-132570780 on Build GRCh38
rs756883952 Chr.6: 132570769-132570769 on Build GRCh38
rs779798582 Chr.6: 132570843-132570843 on Build GRCh38
rs17061401 Chr.6: 132570814-132570814 on Build GRCh38
rs770655651 Chr.6: 132570653-132570653 on Build GRCh38
rs41298397 Chr.6: 132570838-132570838 on Build GRCh38
rs777729106 Chr.6: 132570680-132570680 on Build GRCh38
rs374014146 Chr.6: 132570675-132570675 on Build GRCh38
rs773313599 Chr.6: 132570736-132570736 on Build GRCh38
rs768716058 Chr.6: 132570733-132570734 on Build GRCh38
rs753892369 Chr.6: 132570826-132570826 on Build GRCh38
rs17061404 Chr.6: 132570839-132570839 on Build GRCh38
rs759694783 Chr.6: 132570802-132570802 on Build GRCh38
rs781433346 Chr.6: 132570781-132570781 on Build GRCh38

View Full Product Details