Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 96240743-96241002 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BDKRB2
Entrez Gene ID
624
Gene Name
bradykinin receptor B2
Gene Aliases
B2R, BK-2, BK2, BKR2, BRB2
UniGene
Hs.654542
Gene Chromosome Location
Chr.14: 96204798-96244329 on Build GRCh38
Transcripts
NM_000623.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4504036 c.654C>T p.L218L Chr.14: 96240982-96240982 on Build GRCh38
699706 c.580G>T p.V194L Chr.14: 96240908-96240908 on Build GRCh38
699708 c.579G>T p.L193L Chr.14: 96240907-96240907 on Build GRCh38
1749054 c.515G>A p.R172H Chr.14: 96240843-96240843 on Build GRCh38
3499554 c.492C>T p.S164S Chr.14: 96240820-96240820 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758629002 Chr.14: 96240770-96240770 on Build GRCh38
rs140161378 Chr.14: 96240842-96240842 on Build GRCh38
rs749275929 Chr.14: 96240882-96240882 on Build GRCh38
rs546829915 Chr.14: 96240908-96240908 on Build GRCh38
rs994156703 Chr.14: 96240952-96240952 on Build GRCh38
rs139401938 Chr.14: 96240839-96240839 on Build GRCh38
rs779141421 Chr.14: 96240766-96240766 on Build GRCh38
rs751762638 Chr.14: 96240969-96240969 on Build GRCh38
rs200470118 Chr.14: 96240949-96240949 on Build GRCh38
rs202134620 Chr.14: 96240836-96240836 on Build GRCh38
rs768529413 Chr.14: 96240801-96240801 on Build GRCh38
rs977838309 Chr.14: 96240849-96240849 on Build GRCh38
rs780168746 Chr.14: 96240874-96240874 on Build GRCh38
rs199642464 Chr.14: 96240859-96240859 on Build GRCh38
rs747911761 Chr.14: 96240913-96240913 on Build GRCh38
rs139203012 Chr.14: 96240950-96240950 on Build GRCh38
rs201474162 Chr.14: 96240843-96240843 on Build GRCh38
rs964993900 Chr.14: 96240831-96240831 on Build GRCh38
rs757519944 Chr.14: 96240972-96240972 on Build GRCh38
rs747103685 Chr.14: 96240781-96240781 on Build GRCh38
rs780703098 Chr.14: 96240934-96240934 on Build GRCh38
rs150911844 Chr.14: 96240835-96240835 on Build GRCh38
rs201148125 Chr.14: 96240967-96240967 on Build GRCh38
rs147247250 Chr.14: 96240814-96240814 on Build GRCh38
rs199831344 Chr.14: 96240925-96240925 on Build GRCh38
rs781468888 Chr.14: 96240980-96240980 on Build GRCh38
rs199803197 Chr.14: 96240962-96240962 on Build GRCh38
rs749114824 Chr.14: 96240790-96240790 on Build GRCh38
rs772874206 Chr.14: 96240935-96240935 on Build GRCh38
rs777777245 Chr.14: 96240774-96240774 on Build GRCh38
rs200839127 Chr.14: 96240773-96240773 on Build GRCh38
rs200159176 Chr.14: 96240838-96240838 on Build GRCh38
rs201760673 Chr.14: 96240915-96240915 on Build GRCh38
rs200062156 Chr.14: 96240954-96240954 on Build GRCh38
rs962382149 Chr.14: 96240914-96240914 on Build GRCh38
rs923683046 Chr.14: 96240877-96240877 on Build GRCh38
rs141412276 Chr.14: 96240834-96240834 on Build GRCh38
rs139076079 Chr.14: 96240832-96240832 on Build GRCh38
rs199722917 Chr.14: 96240841-96240841 on Build GRCh38
rs781770613 Chr.14: 96240768-96240768 on Build GRCh38
rs183204823 Chr.14: 96240956-96240956 on Build GRCh38
rs201329154 Chr.14: 96240840-96240840 on Build GRCh38
rs748411805 Chr.14: 96240769-96240769 on Build GRCh38
rs201523214 Chr.14: 96240782-96240782 on Build GRCh38
rs769618803 Chr.14: 96240786-96240786 on Build GRCh38
rs774254602 Chr.14: 96240827-96240827 on Build GRCh38
rs760136048 Chr.14: 96240944-96240944 on Build GRCh38
rs777611379 Chr.14: 96240788-96240788 on Build GRCh38
rs767439867 Chr.14: 96240894-96240894 on Build GRCh38
rs142406520 Chr.14: 96240883-96240883 on Build GRCh38
rs939563124 Chr.14: 96240879-96240879 on Build GRCh38
rs201268118 Chr.14: 96240955-96240955 on Build GRCh38
rs775508034 Chr.14: 96240787-96240787 on Build GRCh38
rs201092575 Chr.14: 96240828-96240828 on Build GRCh38
rs200137610 Chr.14: 96240822-96240822 on Build GRCh38

View Full Product Details