Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 70055040-70055287 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TLE3
Entrez Gene ID
7090
Gene Name
transducin like enhancer of split 3
Gene Aliases
ESG, ESG3, GRG3, HsT18976
UniGene
Hs.287362
Gene Chromosome Location
Chr.15: 70047790-70097935 on Build GRCh38
Transcripts
NM_001105192.2 NM_001282979.1 NM_001282980.1 NM_001282981.1 NM_001282982.1 NM_005078.3 NM_020908.2 XM_005254622.4 XM_005254623.3 XM_005254625.3 XM_005254628.3 XM_005254633.2 XM_006720665.3 XM_011521976.2 XM_011521977.2 XM_011521978.2 XM_011521979.2 XM_011521980.2 XM_011521981.1 XM_011521982.2 XM_011521983.2 XM_017022532.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1189052 c.1399G>T p.A467S Chr.15: 70055237-70055237 on Build GRCh38
7288666 c.1549C>T p.P517S Chr.15: 70055087-70055087 on Build GRCh38
6802222 c.1522T>C p.C508R Chr.15: 70055114-70055114 on Build GRCh38
5524009 c.1420C>T p.P474S Chr.15: 70055216-70055216 on Build GRCh38
6802226 c.1399G>A p.A467T Chr.15: 70055237-70055237 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766356098 Chr.15: 70055204-70055204 on Build GRCh38
rs201791846 Chr.15: 70055214-70055214 on Build GRCh38
rs751485225 Chr.15: 70055070-70055070 on Build GRCh38
rs776949492 Chr.15: 70055169-70055169 on Build GRCh38
rs201049660 Chr.15: 70055238-70055238 on Build GRCh38
rs200543589 Chr.15: 70055178-70055178 on Build GRCh38
rs535644010 Chr.15: 70055188-70055188 on Build GRCh38
rs773242711 Chr.15: 70055192-70055192 on Build GRCh38
rs368231867 Chr.15: 70055142-70055142 on Build GRCh38
rs747553373 Chr.15: 70055243-70055243 on Build GRCh38
rs1046998233 Chr.15: 70055187-70055187 on Build GRCh38
rs767474312 Chr.15: 70055212-70055212 on Build GRCh38
rs763110373 Chr.15: 70055203-70055203 on Build GRCh38
rs374848525 Chr.15: 70055111-70055111 on Build GRCh38
rs751614749 Chr.15: 70055207-70055207 on Build GRCh38
rs554160023 Chr.15: 70055244-70055244 on Build GRCh38
rs7179730 Chr.15: 70055136-70055136 on Build GRCh38
rs747498036 Chr.15: 70055163-70055163 on Build GRCh38
rs370946243 Chr.15: 70055223-70055223 on Build GRCh38
rs948206486 Chr.15: 70055180-70055180 on Build GRCh38
rs748831377 Chr.15: 70055225-70055225 on Build GRCh38
rs200517303 Chr.15: 70055256-70055256 on Build GRCh38
rs982632223 Chr.15: 70055076-70055076 on Build GRCh38
rs748518029 Chr.15: 70055247-70055247 on Build GRCh38
rs754878560 Chr.15: 70055073-70055073 on Build GRCh38
rs748858713 Chr.15: 70055139-70055139 on Build GRCh38
rs754938695 Chr.15: 70055208-70055208 on Build GRCh38
rs769997767 Chr.15: 70055250-70055250 on Build GRCh38
rs747847895 Chr.15: 70055097-70055097 on Build GRCh38
rs762068389 Chr.15: 70055181-70055181 on Build GRCh38
rs893233239 Chr.15: 70055211-70055211 on Build GRCh38
rs755829203 Chr.15: 70055222-70055222 on Build GRCh38
rs756844919 Chr.15: 70055228-70055228 on Build GRCh38
rs781133639 Chr.15: 70055095-70055095 on Build GRCh38
rs1055690023 Chr.15: 70055189-70055189 on Build GRCh38
rs200508723 Chr.15: 70055143-70055143 on Build GRCh38

View Full Product Details