Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 53978926-53979199 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TCEA1
Entrez Gene ID
6917
Gene Name
transcription elongation factor A1
Gene Aliases
GTF2S, SII, TCEA, TF2S, TFIIS
UniGene
Hs.491745
Gene Chromosome Location
Chr.8: 53966554-54022456 on Build GRCh38
Transcripts
NM_006756.3 NM_201437.2 NR_109901.1 NR_109902.1 XM_006716467.1 XM_011517578.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4168192 c.764G>T p.G255V Chr.8: 53979086-53979086 on Build GRCh38
6304739 c.231C>A p.F77L Chr.8: 53979067-53979067 on Build GRCh38
230626 c.185G>A p.R62K Chr.8: 53979113-53979113 on Build GRCh38
6217335 c.127-1G>A p.? Chr.8: 53979172-53979172 on Build GRCh38
5184389 c.787T>C p.C263R Chr.8: 53979063-53979063 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759228331 Chr.8: 53979044-53979044 on Build GRCh38
rs758708704 Chr.8: 53979136-53979136 on Build GRCh38
rs767481992 Chr.8: 53979069-53979069 on Build GRCh38
rs779033279 Chr.8: 53978964-53978964 on Build GRCh38
rs757344506 Chr.8: 53979124-53979124 on Build GRCh38
rs965053030 Chr.8: 53979178-53979178 on Build GRCh38
rs769377427 Chr.8: 53978984-53978984 on Build GRCh38
rs773009388 Chr.8: 53979003-53979003 on Build GRCh38
rs769259768 Chr.8: 53978976-53978976 on Build GRCh38
rs751428994 Chr.8: 53979041-53979041 on Build GRCh38
rs200499230 Chr.8: 53979140-53979140 on Build GRCh38
rs374150495 Chr.8: 53979064-53979064 on Build GRCh38
rs977786885 Chr.8: 53979163-53979163 on Build GRCh38
rs376368525 Chr.8: 53978982-53978982 on Build GRCh38
rs778897877 Chr.8: 53979127-53979127 on Build GRCh38
rs965028555 Chr.8: 53979095-53979095 on Build GRCh38
rs1026861475 Chr.8: 53979008-53979008 on Build GRCh38
rs371090622 Chr.8: 53979031-53979031 on Build GRCh38
rs772596271 Chr.8: 53979143-53979143 on Build GRCh38
rs781346211 Chr.8: 53979003-53979004 on Build GRCh38
rs777248974 Chr.8: 53978979-53978979 on Build GRCh38
rs756311513 Chr.8: 53979019-53979019 on Build GRCh38
rs368515547 Chr.8: 53979090-53979090 on Build GRCh38
rs762271675 Chr.8: 53979108-53979108 on Build GRCh38
rs535625630 Chr.8: 53978994-53978994 on Build GRCh38
rs750465931 Chr.8: 53979134-53979134 on Build GRCh38
rs201331730 Chr.8: 53979174-53979174 on Build GRCh38
rs954248322 Chr.8: 53978967-53978967 on Build GRCh38

View Full Product Details