Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 70689364-70689601 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UTP3
Entrez Gene ID
57050
Gene Name
UTP3, small subunit processome component homolog (S. cerevisiae)
Gene Aliases
CRL1, CRLZ1, SAS10
UniGene
Hs.322901
Gene Chromosome Location
Chr.4: 70688479-70690551 on Build GRCh38
Transcripts
NM_020368.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5443543 c.750G>C p.L250F Chr.4: 70689427-70689427 on Build GRCh38
4125766 c.764T>A p.I255N Chr.4: 70689441-70689441 on Build GRCh38
7144268 c.855G>T p.L285L Chr.4: 70689532-70689532 on Build GRCh38
7144269 c.864G>C p.R288S Chr.4: 70689541-70689541 on Build GRCh38
7192344 c.821A>G p.N274S Chr.4: 70689498-70689498 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146828085 Chr.4: 70689547-70689547 on Build GRCh38
rs773843356 Chr.4: 70689516-70689516 on Build GRCh38
rs772496914 Chr.4: 70689436-70689436 on Build GRCh38
rs571545362 Chr.4: 70689451-70689451 on Build GRCh38
rs995644939 Chr.4: 70689494-70689494 on Build GRCh38
rs763993235 Chr.4: 70689505-70689505 on Build GRCh38
rs751491812 Chr.4: 70689511-70689511 on Build GRCh38
rs779165784 Chr.4: 70689567-70689567 on Build GRCh38
rs141537804 Chr.4: 70689417-70689417 on Build GRCh38
rs1025324042 Chr.4: 70689521-70689521 on Build GRCh38
rs757118177 Chr.4: 70689525-70689525 on Build GRCh38
rs780319686 Chr.4: 70689421-70689421 on Build GRCh38
rs370336880 Chr.4: 70689556-70689556 on Build GRCh38
rs764898934 Chr.4: 70689481-70689481 on Build GRCh38
rs150243800 Chr.4: 70689399-70689399 on Build GRCh38
rs759280455 Chr.4: 70689474-70689474 on Build GRCh38
rs1011571259 Chr.4: 70689555-70689555 on Build GRCh38
rs1031574180 Chr.4: 70689400-70689400 on Build GRCh38
rs779166781 Chr.4: 70689434-70689434 on Build GRCh38
rs776217495 Chr.4: 70689459-70689459 on Build GRCh38
rs750054886 Chr.4: 70689555-70689556 on Build GRCh38
rs898271557 Chr.4: 70689466-70689466 on Build GRCh38
rs752471235 Chr.4: 70689488-70689488 on Build GRCh38
rs780120040 Chr.4: 70689540-70689540 on Build GRCh38
rs760321355 Chr.4: 70689449-70689449 on Build GRCh38
rs762783396 Chr.4: 70689502-70689502 on Build GRCh38
rs1002459236 Chr.4: 70689549-70689549 on Build GRCh38
rs749514015 Chr.4: 70689427-70689427 on Build GRCh38
rs773748650 Chr.4: 70689441-70689441 on Build GRCh38
rs764909696 Chr.4: 70689420-70689421 on Build GRCh38
rs141044445 Chr.4: 70689395-70689395 on Build GRCh38

View Full Product Details