Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44081916-44082163 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF284
Entrez Gene ID
342909
Gene Name
zinc finger protein 284
Gene Aliases
ZNF284L
UniGene
Hs.445395
Gene Chromosome Location
Chr.19: 44072137-44089613 on Build GRCh38
Transcripts
NM_001037813.2 XM_011526907.2 XM_011526908.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5707066 c.167C>T p.T56I Chr.19: 44082037-44082037 on Build GRCh38
3535429 c.172C>T p.H58Y Chr.19: 44082042-44082042 on Build GRCh38
3535428 c.156C>T p.S52S Chr.19: 44082026-44082026 on Build GRCh38
4982742 c.184G>A p.E62K Chr.19: 44082054-44082054 on Build GRCh38
4743426 c.188A>C p.E63A Chr.19: 44082058-44082058 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769462357 Chr.19: 44081979-44081979 on Build GRCh38
rs987203514 Chr.19: 44081961-44081961 on Build GRCh38
rs367985070 Chr.19: 44082096-44082096 on Build GRCh38
rs1031006967 Chr.19: 44082104-44082104 on Build GRCh38
rs756341420 Chr.19: 44082048-44082048 on Build GRCh38
rs772869984 Chr.19: 44082106-44082106 on Build GRCh38
rs193163502 Chr.19: 44082067-44082067 on Build GRCh38
rs767649596 Chr.19: 44082018-44082018 on Build GRCh38
rs202186808 Chr.19: 44082016-44082016 on Build GRCh38
rs773486603 Chr.19: 44081982-44081982 on Build GRCh38
rs371971611 Chr.19: 44082133-44082133 on Build GRCh38
rs538798803 Chr.19: 44082098-44082098 on Build GRCh38
rs1012398082 Chr.19: 44081941-44081941 on Build GRCh38
rs771216276 Chr.19: 44082125-44082125 on Build GRCh38
rs774694237 Chr.19: 44082132-44082132 on Build GRCh38
rs993734681 Chr.19: 44082127-44082127 on Build GRCh38
rs374076216 Chr.19: 44082075-44082075 on Build GRCh38
rs763240160 Chr.19: 44082121-44082121 on Build GRCh38
rs201560410 Chr.19: 44082061-44082061 on Build GRCh38
rs746879511 Chr.19: 44082089-44082089 on Build GRCh38
rs764506939 Chr.19: 44081990-44081991 on Build GRCh38
rs766813463 Chr.19: 44081984-44081984 on Build GRCh38
rs778027999 Chr.19: 44082059-44082059 on Build GRCh38
rs368672902 Chr.19: 44082137-44082137 on Build GRCh38
rs963984316 Chr.19: 44082122-44082122 on Build GRCh38
rs778358399 Chr.19: 44082074-44082074 on Build GRCh38
rs769360604 Chr.19: 44082099-44082099 on Build GRCh38
rs780158340 Chr.19: 44082080-44082080 on Build GRCh38
rs752760034 Chr.19: 44082029-44082029 on Build GRCh38
rs371661208 Chr.19: 44081991-44081992 on Build GRCh38
rs761628141 Chr.19: 44081974-44081974 on Build GRCh38
rs1046462195 Chr.19: 44082031-44082031 on Build GRCh38
rs200294399 Chr.19: 44082072-44082072 on Build GRCh38
rs754162915 Chr.19: 44082124-44082124 on Build GRCh38
rs1002623252 Chr.19: 44082054-44082054 on Build GRCh38
rs375112419 Chr.19: 44082136-44082136 on Build GRCh38
rs910241301 Chr.19: 44081970-44081970 on Build GRCh38
rs752005375 Chr.19: 44081995-44081995 on Build GRCh38
rs1024131492 Chr.19: 44082004-44082004 on Build GRCh38
rs750216634 Chr.19: 44082073-44082073 on Build GRCh38

View Full Product Details