Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 36089018-36089259 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADPRHL2
Entrez Gene ID
54936
Gene Name
ADP-ribosylhydrolase like 2
Gene Aliases
ARH3
UniGene
Hs.18021
Gene Chromosome Location
Chr.1: 36088852-36093932 on Build GRCh38
Transcripts
NM_017825.2 XM_011541636.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6408686 c.190A>G p.T64A Chr.1: 36089094-36089094 on Build GRCh38
5799603 c.121G>T p.E41* Chr.1: 36089025-36089025 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs781657776 Chr.1: 36089085-36089085 on Build GRCh38
rs918110783 Chr.1: 36089206-36089206 on Build GRCh38
rs773923767 Chr.1: 36089132-36089132 on Build GRCh38
rs948261259 Chr.1: 36089220-36089220 on Build GRCh38
rs965644152 Chr.1: 36089203-36089203 on Build GRCh38
rs191037069 Chr.1: 36089236-36089236 on Build GRCh38
rs773644447 Chr.1: 36089110-36089110 on Build GRCh38
rs1013852896 Chr.1: 36089118-36089119 on Build GRCh38
rs1021518861 Chr.1: 36089121-36089121 on Build GRCh38
rs747084873 Chr.1: 36089081-36089081 on Build GRCh38
rs759072063 Chr.1: 36089149-36089149 on Build GRCh38
rs982418864 Chr.1: 36089146-36089146 on Build GRCh38
rs755103060 Chr.1: 36089083-36089083 on Build GRCh38
rs912256602 Chr.1: 36089182-36089182 on Build GRCh38
rs368043338 Chr.1: 36089061-36089061 on Build GRCh38
rs775503829 Chr.1: 36089159-36089159 on Build GRCh38
rs187776843 Chr.1: 36089090-36089090 on Build GRCh38
rs779962601 Chr.1: 36089069-36089069 on Build GRCh38
rs748847636 Chr.1: 36089127-36089127 on Build GRCh38
rs557002734 Chr.1: 36089214-36089214 on Build GRCh38
rs759007491 Chr.1: 36089039-36089039 on Build GRCh38
rs750553450 Chr.1: 36089060-36089060 on Build GRCh38
rs763988269 Chr.1: 36089057-36089057 on Build GRCh38
rs963899157 Chr.1: 36089107-36089107 on Build GRCh38
rs745658604 Chr.1: 36089139-36089140 on Build GRCh38
rs866188444 Chr.1: 36089062-36089062 on Build GRCh38
rs537091313 Chr.1: 36089239-36089239 on Build GRCh38
rs752738366 Chr.1: 36089043-36089043 on Build GRCh38
rs770585649 Chr.1: 36089128-36089128 on Build GRCh38
rs952423952 Chr.1: 36089145-36089145 on Build GRCh38
rs749889837 Chr.1: 36089040-36089040 on Build GRCh38
rs760704035 Chr.1: 36089161-36089161 on Build GRCh38
rs755967272 Chr.1: 36089050-36089050 on Build GRCh38
rs867567402 Chr.1: 36089109-36089109 on Build GRCh38
rs200675752 Chr.1: 36089136-36089136 on Build GRCh38
rs887896172 Chr.1: 36089090-36089094 on Build GRCh38
rs758137622 Chr.1: 36089219-36089219 on Build GRCh38
rs770083637 Chr.1: 36089100-36089100 on Build GRCh38
rs972693495 Chr.1: 36089205-36089205 on Build GRCh38
rs1033704731 Chr.1: 36089096-36089096 on Build GRCh38
rs372297585 Chr.1: 36089190-36089190 on Build GRCh38
rs758669840 Chr.1: 36089066-36089066 on Build GRCh38
rs535622654 Chr.1: 36089172-36089172 on Build GRCh38

View Full Product Details