Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1499156-1499409 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TELO2
Entrez Gene ID
9894
Gene Name
telomere maintenance 2
Gene Aliases
CLK2, TEL2, YHFS
UniGene
Hs.271044
Gene Chromosome Location
Chr.16: 1493344-1510459 on Build GRCh38
Transcripts
NM_016111.3 XM_011522773.2 XM_011522774.2 XM_011522775.2 XM_011522776.2 XM_011522777.2 XM_011522778.2 XM_017023914.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4058514 c.887A>C p.Q296P Chr.16: 1499287-1499287 on Build GRCh38
118778 c.864G>A p.L288L Chr.16: 1499264-1499264 on Build GRCh38
334016 c.920_921AG>TA p.Q307>? Chr.16: 1499320-1499321 on Build GRCh38
5850223 c.929T>A p.L310H Chr.16: 1499329-1499329 on Build GRCh38
4426227 c.925C>T p.R309W Chr.16: 1499325-1499325 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370018942 Chr.16: 1499291-1499291 on Build GRCh38
rs937115852 Chr.16: 1499355-1499356 on Build GRCh38
rs201783779 Chr.16: 1499354-1499354 on Build GRCh38
rs745439594 Chr.16: 1499342-1499342 on Build GRCh38
rs144312433 Chr.16: 1499326-1499326 on Build GRCh38
rs780740357 Chr.16: 1499345-1499345 on Build GRCh38
rs766757086 Chr.16: 1499299-1499299 on Build GRCh38
rs762337620 Chr.16: 1499261-1499261 on Build GRCh38
rs769185024 Chr.16: 1499347-1499347 on Build GRCh38
rs753958889 Chr.16: 1499303-1499303 on Build GRCh38
rs773743489 Chr.16: 1499366-1499366 on Build GRCh38
rs760582253 Chr.16: 1499205-1499205 on Build GRCh38
rs758900311 Chr.16: 1499243-1499243 on Build GRCh38
rs778937429 Chr.16: 1499195-1499195 on Build GRCh38
rs763794046 Chr.16: 1499363-1499363 on Build GRCh38
rs771847745 Chr.16: 1499202-1499202 on Build GRCh38
rs779259878 Chr.16: 1499313-1499313 on Build GRCh38
rs765276863 Chr.16: 1499372-1499372 on Build GRCh38
rs764952387 Chr.16: 1499246-1499246 on Build GRCh38
rs377524934 Chr.16: 1499380-1499380 on Build GRCh38
rs578067307 Chr.16: 1499375-1499375 on Build GRCh38
rs780163740 Chr.16: 1499189-1499189 on Build GRCh38
rs767967393 Chr.16: 1499264-1499264 on Build GRCh38
rs775266511 Chr.16: 1499250-1499250 on Build GRCh38
rs200020791 Chr.16: 1499188-1499188 on Build GRCh38
rs11344586 Chr.16: 1499343-1499343 on Build GRCh38
rs766923402 Chr.16: 1499370-1499370 on Build GRCh38
rs758238068 Chr.16: 1499323-1499323 on Build GRCh38
rs540233352 Chr.16: 1499384-1499384 on Build GRCh38
rs747951919 Chr.16: 1499196-1499196 on Build GRCh38
rs947224120 Chr.16: 1499379-1499379 on Build GRCh38
rs775325852 Chr.16: 1499350-1499350 on Build GRCh38
rs142640113 Chr.16: 1499310-1499310 on Build GRCh38
rs201883234 Chr.16: 1499191-1499191 on Build GRCh38
rs113219709 Chr.16: 1499182-1499182 on Build GRCh38
rs985542325 Chr.16: 1499219-1499219 on Build GRCh38
rs751021623 Chr.16: 1499277-1499277 on Build GRCh38
rs749278986 Chr.16: 1499190-1499190 on Build GRCh38
rs776371581 Chr.16: 1499341-1499341 on Build GRCh38
rs760882962 Chr.16: 1499369-1499369 on Build GRCh38
rs373899324 Chr.16: 1499325-1499325 on Build GRCh38
rs903107423 Chr.16: 1499381-1499381 on Build GRCh38
rs200185348 Chr.16: 1499332-1499332 on Build GRCh38
rs373799263 Chr.16: 1499285-1499285 on Build GRCh38
rs370808477 Chr.16: 1499318-1499318 on Build GRCh38
rs770724067 Chr.16: 1499231-1499231 on Build GRCh38
rs755123206 Chr.16: 1499371-1499371 on Build GRCh38
rs148740663 Chr.16: 1499232-1499232 on Build GRCh38
rs773172247 Chr.16: 1499204-1499204 on Build GRCh38
rs936416087 Chr.16: 1499346-1499346 on Build GRCh38
rs397854665 Chr.16: 1499344-1499344 on Build GRCh38

View Full Product Details