Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 160049549-160049774 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PWWP2A
Entrez Gene ID
114825
Gene Name
PWWP domain containing 2A
Gene Aliases
MST101
UniGene
Hs.744960
Gene Chromosome Location
Chr.5: 160043538-160120709 on Build GRCh38
Transcripts
XM_011534424.2 NM_001282500.1 NM_003314.2

Gene Symbol
TTC1
Entrez Gene ID
7265
Gene Name
tetratricopeptide repeat domain 1
Gene Aliases
TPR1
UniGene
Hs.519718
Gene Chromosome Location
Chr.5: 160009100-160065545 on Build GRCh38
Transcripts
XM_011534424.2 NM_001282500.1 NM_003314.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4504201 c.658C>T p.P220S Chr.5: 160049630-160049630 on Build GRCh38
5756620 c.605C>T p.T202M Chr.5: 160049577-160049577 on Build GRCh38
165217 c.683C>T p.A228V Chr.5: 160049655-160049655 on Build GRCh38
4155585 c.669T>G p.H223Q Chr.5: 160049641-160049641 on Build GRCh38
217356 c.635A>G p.Y212C Chr.5: 160049607-160049607 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747295685 Chr.5: 160049641-160049641 on Build GRCh38
rs554395265 Chr.5: 160049577-160049577 on Build GRCh38
rs373880865 Chr.5: 160049710-160049710 on Build GRCh38
rs376379489 Chr.5: 160049636-160049636 on Build GRCh38
rs748728919 Chr.5: 160049670-160049670 on Build GRCh38
rs991214506 Chr.5: 160049653-160049653 on Build GRCh38
rs773768730 Chr.5: 160049693-160049693 on Build GRCh38
rs370906460 Chr.5: 160049707-160049707 on Build GRCh38
rs751388000 Chr.5: 160049649-160049650 on Build GRCh38
rs759182630 Chr.5: 160049698-160049698 on Build GRCh38
rs752512742 Chr.5: 160049607-160049607 on Build GRCh38
rs570764437 Chr.5: 160049578-160049578 on Build GRCh38
rs773919436 Chr.5: 160049665-160049665 on Build GRCh38
rs377300902 Chr.5: 160049678-160049678 on Build GRCh38
rs755988160 Chr.5: 160049615-160049615 on Build GRCh38
rs1054861870 Chr.5: 160049718-160049718 on Build GRCh38
rs928509674 Chr.5: 160049582-160049582 on Build GRCh38
rs182036182 Chr.5: 160049701-160049701 on Build GRCh38
rs748533174 Chr.5: 160049655-160049655 on Build GRCh38
rs758562953 Chr.5: 160049639-160049639 on Build GRCh38
rs780115205 Chr.5: 160049640-160049640 on Build GRCh38
rs756939767 Chr.5: 160049702-160049702 on Build GRCh38
rs759224520 Chr.5: 160049588-160049588 on Build GRCh38
rs768790565 Chr.5: 160049642-160049642 on Build GRCh38
rs767263458 Chr.5: 160049597-160049597 on Build GRCh38
rs151026006 Chr.5: 160049619-160049619 on Build GRCh38
rs370202062 Chr.5: 160049649-160049649 on Build GRCh38
rs893424413 Chr.5: 160049733-160049733 on Build GRCh38
rs373530006 Chr.5: 160049606-160049606 on Build GRCh38
rs770355249 Chr.5: 160049660-160049660 on Build GRCh38

View Full Product Details