Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 57872646-57872901 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF814
Entrez Gene ID
730051
Gene Name
zinc finger protein 814
Gene Aliases
-
UniGene
Hs.634143
Gene Chromosome Location
Chr.19: 57869379-57889074 on Build GRCh38
Transcripts
NM_001144989.1 XM_017027197.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7199732 c.2565_2566delAT p.I855fs? Chr.19: 57872824-57872825 on Build GRCh38
5464804 c.2560G>A p.A854T Chr.19: 57872830-57872830 on Build GRCh38
1612971 c.2503A>C p.K835Q Chr.19: 57872887-57872887 on Build GRCh38
5605482 c.2508A>G p.L836L Chr.19: 57872882-57872882 on Build GRCh38
1002505 c.2505delA p.K835fs*>21 Chr.19: 57872885-57872885 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1041784573 Chr.19: 57872785-57872785 on Build GRCh38
rs1047047641 Chr.19: 57872741-57872741 on Build GRCh38
rs759485120 Chr.19: 57872864-57872864 on Build GRCh38
rs761697430 Chr.19: 57872805-57872806 on Build GRCh38
rs376099860 Chr.19: 57872835-57872835 on Build GRCh38
rs907932399 Chr.19: 57872720-57872720 on Build GRCh38
rs750583122 Chr.19: 57872797-57872797 on Build GRCh38
rs1037817856 Chr.19: 57872730-57872730 on Build GRCh38
rs1052634637 Chr.19: 57872808-57872808 on Build GRCh38
rs562254756 Chr.19: 57872716-57872716 on Build GRCh38
rs987456968 Chr.19: 57872745-57872745 on Build GRCh38
rs760789890 Chr.19: 57872766-57872766 on Build GRCh38
rs932277846 Chr.19: 57872743-57872743 on Build GRCh38
rs973633287 Chr.19: 57872733-57872733 on Build GRCh38
rs780053139 Chr.19: 57872807-57872807 on Build GRCh38
rs756192498 Chr.19: 57872803-57872803 on Build GRCh38
rs777671410 Chr.19: 57872824-57872824 on Build GRCh38
rs983138212 Chr.19: 57872711-57872711 on Build GRCh38
rs768834125 Chr.19: 57872820-57872820 on Build GRCh38
rs946049315 Chr.19: 57872781-57872781 on Build GRCh38
rs149535034 Chr.19: 57872726-57872726 on Build GRCh38
rs751842928 Chr.19: 57872783-57872783 on Build GRCh38
rs970378644 Chr.19: 57872679-57872679 on Build GRCh38
rs749536496 Chr.19: 57872819-57872819 on Build GRCh38
rs936498485 Chr.19: 57872822-57872822 on Build GRCh38
rs776432920 Chr.19: 57872863-57872863 on Build GRCh38
rs764587286 Chr.19: 57872777-57872777 on Build GRCh38
rs533402715 Chr.19: 57872851-57872851 on Build GRCh38
rs752922935 Chr.19: 57872736-57872736 on Build GRCh38
rs541447664 Chr.19: 57872748-57872748 on Build GRCh38
rs781541818 Chr.19: 57872795-57872795 on Build GRCh38
rs144091647 Chr.19: 57872792-57872792 on Build GRCh38

View Full Product Details