Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 46352178-46352407 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC35C2
Entrez Gene ID
51006
Gene Name
solute carrier family 35 member C2
Gene Aliases
BA394O2.1, C20orf5, CGI-15, OVCOV1
UniGene
Hs.593344
Gene Chromosome Location
Chr.20: 46349528-46364461 on Build GRCh38
Transcripts
NM_001281457.1 NM_001281458.1 NM_001281459.1 NM_001281460.1 NM_015945.11 NM_173073.3 NM_173179.3 XM_011528831.1 XM_011528832.1 XM_011528833.1 XM_011528834.1 XM_011528835.1 XM_011528836.1 XM_011528837.1 XM_011528838.1 XM_017027856.1 XM_017027857.1 XM_017027858.1 XM_017027859.1 XM_017027860.1 XM_017027861.1 XM_017027862.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4821631 c.281C>G p.S94C Chr.20: 46352208-46352208 on Build GRCh38
6781665 c.795A>C p.T265T Chr.20: 46352180-46352180 on Build GRCh38
4821630 c.767C>G p.S256C Chr.20: 46352208-46352208 on Build GRCh38
5038068 c.760T>G p.S254A Chr.20: 46352215-46352215 on Build GRCh38
6781664 c.708A>C p.T236T Chr.20: 46352180-46352180 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs932295465 Chr.20: 46352374-46352374 on Build GRCh38
rs751943659 Chr.20: 46352237-46352238 on Build GRCh38
rs375805673 Chr.20: 46352277-46352277 on Build GRCh38
rs536422983 Chr.20: 46352301-46352301 on Build GRCh38
rs538891132 Chr.20: 46352241-46352241 on Build GRCh38
rs770675016 Chr.20: 46352240-46352240 on Build GRCh38
rs575314359 Chr.20: 46352292-46352292 on Build GRCh38
rs965108384 Chr.20: 46352311-46352311 on Build GRCh38
rs745855673 Chr.20: 46352247-46352247 on Build GRCh38
rs372872421 Chr.20: 46352258-46352258 on Build GRCh38
rs555318746 Chr.20: 46352359-46352359 on Build GRCh38
rs776304697 Chr.20: 46352265-46352265 on Build GRCh38
rs749131022 Chr.20: 46352222-46352222 on Build GRCh38
rs755267743 Chr.20: 46352214-46352214 on Build GRCh38
rs777665689 Chr.20: 46352219-46352219 on Build GRCh38
rs762215200 Chr.20: 46352268-46352268 on Build GRCh38
rs557247845 Chr.20: 46352253-46352253 on Build GRCh38
rs952601733 Chr.20: 46352304-46352304 on Build GRCh38
rs908335028 Chr.20: 46352310-46352310 on Build GRCh38
rs760269620 Chr.20: 46352256-46352256 on Build GRCh38
rs997354120 Chr.20: 46352302-46352302 on Build GRCh38
rs570005796 Chr.20: 46352357-46352357 on Build GRCh38
rs774990946 Chr.20: 46352249-46352249 on Build GRCh38
rs750082853 Chr.20: 46352349-46352351 on Build GRCh38
rs1022973124 Chr.20: 46352210-46352210 on Build GRCh38
rs764674736 Chr.20: 46352234-46352234 on Build GRCh38
rs1043338394 Chr.20: 46352381-46352381 on Build GRCh38
rs771541901 Chr.20: 46352248-46352248 on Build GRCh38
rs763303679 Chr.20: 46352199-46352200 on Build GRCh38
rs745463054 Chr.20: 46352313-46352313 on Build GRCh38
rs1007326340 Chr.20: 46352372-46352372 on Build GRCh38
rs751765985 Chr.20: 46352200-46352200 on Build GRCh38
rs982577421 Chr.20: 46352309-46352309 on Build GRCh38
rs572989050 Chr.20: 46352368-46352368 on Build GRCh38
rs540337203 Chr.20: 46352388-46352388 on Build GRCh38
rs757388729 Chr.20: 46352251-46352252 on Build GRCh38

View Full Product Details