Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 129167670-129167937 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSPAN33
Entrez Gene ID
340348
Gene Name
tetraspanin 33
Gene Aliases
PEN, PEN., TSPAN-33
UniGene
Hs.27267
Gene Chromosome Location
Chr.7: 129144716-129169694 on Build GRCh38
Transcripts
NM_178562.4 XM_006715960.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5871796 c.787C>T p.Q263* Chr.7: 129167809-129167809 on Build GRCh38
2769691 c.829C>T p.H277Y Chr.7: 129167851-129167851 on Build GRCh38
6530255 c.796G>C p.D266H Chr.7: 129167818-129167818 on Build GRCh38
1699345 c.799C>T p.Q267* Chr.7: 129167821-129167821 on Build GRCh38
6810647 c.832C>T p.R278W Chr.7: 129167854-129167854 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143816378 Chr.7: 129167868-129167868 on Build GRCh38
rs963827581 Chr.7: 129167895-129167895 on Build GRCh38
rs951173232 Chr.7: 129167906-129167906 on Build GRCh38
rs751920564 Chr.7: 129167871-129167871 on Build GRCh38
rs753173552 Chr.7: 129167767-129167767 on Build GRCh38
rs771997909 Chr.7: 129167914-129167914 on Build GRCh38
rs949389479 Chr.7: 129167696-129167696 on Build GRCh38
rs768211292 Chr.7: 129167785-129167785 on Build GRCh38
rs377133451 Chr.7: 129167742-129167742 on Build GRCh38
rs563054597 Chr.7: 129167709-129167709 on Build GRCh38
rs779890241 Chr.7: 129167888-129167888 on Build GRCh38
rs776804030 Chr.7: 129167842-129167842 on Build GRCh38
rs748065568 Chr.7: 129167817-129167817 on Build GRCh38
rs759676812 Chr.7: 129167853-129167853 on Build GRCh38
rs148833581 Chr.7: 129167787-129167787 on Build GRCh38
rs765345091 Chr.7: 129167854-129167854 on Build GRCh38
rs35331287 Chr.7: 129167891-129167891 on Build GRCh38
rs762580147 Chr.7: 129167835-129167835 on Build GRCh38
rs755783076 Chr.7: 129167741-129167741 on Build GRCh38
rs763449599 Chr.7: 129167864-129167864 on Build GRCh38
rs753157658 Chr.7: 129167855-129167855 on Build GRCh38
rs771772403 Chr.7: 129167823-129167823 on Build GRCh38
rs772951670 Chr.7: 129167830-129167830 on Build GRCh38
rs779181055 Chr.7: 129167748-129167748 on Build GRCh38
rs73230567 Chr.7: 129167729-129167729 on Build GRCh38
rs750101051 Chr.7: 129167731-129167731 on Build GRCh38
rs148167479 Chr.7: 129167875-129167875 on Build GRCh38
rs753241938 Chr.7: 129167723-129167723 on Build GRCh38
rs753530656 Chr.7: 129167889-129167889 on Build GRCh38
rs754462904 Chr.7: 129167890-129167890 on Build GRCh38
rs748854019 Chr.7: 129167743-129167743 on Build GRCh38
rs765642801 Chr.7: 129167695-129167695 on Build GRCh38
rs750092221 Chr.7: 129167800-129167800 on Build GRCh38
rs894094014 Chr.7: 129167821-129167821 on Build GRCh38
rs771193677 Chr.7: 129167837-129167837 on Build GRCh38

View Full Product Details