Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 64634037-64634288 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACHD1
Entrez Gene ID
57685
Gene Name
cache domain containing 1
Gene Aliases
-
UniGene
Hs.443891
Gene Chromosome Location
Chr.1: 64470793-64693058 on Build GRCh38
Transcripts
NM_001293274.1 NM_020925.2 XM_011541862.1 XM_011541863.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5827955 c.974G>A p.R325Q Chr.1: 64634228-64634228 on Build GRCh38
1343884 c.821G>A p.R274Q Chr.1: 64634228-64634228 on Build GRCh38
2238943 c.687G>A p.Q229Q Chr.1: 64634094-64634094 on Build GRCh38
6317405 c.946G>T p.G316* Chr.1: 64634200-64634200 on Build GRCh38
3491732 c.663C>G p.T221T Chr.1: 64634070-64634070 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs530434842 Chr.1: 64634071-64634071 on Build GRCh38
rs776721588 Chr.1: 64634099-64634099 on Build GRCh38
rs750193456 Chr.1: 64634255-64634255 on Build GRCh38
rs372000913 Chr.1: 64634227-64634227 on Build GRCh38
rs754496507 Chr.1: 64634186-64634186 on Build GRCh38
rs764245231 Chr.1: 64634248-64634248 on Build GRCh38
rs759717842 Chr.1: 64634104-64634104 on Build GRCh38
rs761699943 Chr.1: 64634163-64634163 on Build GRCh38
rs146287870 Chr.1: 64634162-64634162 on Build GRCh38
rs373663142 Chr.1: 64634208-64634208 on Build GRCh38
rs770946584 Chr.1: 64634158-64634160 on Build GRCh38
rs999303032 Chr.1: 64634157-64634157 on Build GRCh38
rs188597931 Chr.1: 64634073-64634073 on Build GRCh38
rs770889563 Chr.1: 64634092-64634092 on Build GRCh38
rs774367782 Chr.1: 64634239-64634239 on Build GRCh38
rs765218619 Chr.1: 64634149-64634149 on Build GRCh38
rs778246703 Chr.1: 64634199-64634199 on Build GRCh38
rs756029885 Chr.1: 64634165-64634165 on Build GRCh38
rs12044872 Chr.1: 64634103-64634103 on Build GRCh38
rs374614547 Chr.1: 64634193-64634193 on Build GRCh38
rs547997798 Chr.1: 64634187-64634187 on Build GRCh38
rs768259419 Chr.1: 64634070-64634070 on Build GRCh38
rs765501518 Chr.1: 64634143-64634143 on Build GRCh38
rs770584445 Chr.1: 64634194-64634194 on Build GRCh38
rs987765201 Chr.1: 64634262-64634262 on Build GRCh38
rs528226784 Chr.1: 64634170-64634170 on Build GRCh38
rs906164297 Chr.1: 64634094-64634094 on Build GRCh38
rs12044869 Chr.1: 64634089-64634089 on Build GRCh38
rs748771715 Chr.1: 64634069-64634069 on Build GRCh38
rs12044863 Chr.1: 64634065-64634065 on Build GRCh38
rs767389262 Chr.1: 64634164-64634164 on Build GRCh38
rs12044906 Chr.1: 64634176-64634176 on Build GRCh38
rs761281548 Chr.1: 64634088-64634088 on Build GRCh38
rs144177105 Chr.1: 64634234-64634234 on Build GRCh38
rs371026074 Chr.1: 64634074-64634074 on Build GRCh38
rs759679081 Chr.1: 64634223-64634223 on Build GRCh38
rs771427370 Chr.1: 64634214-64634214 on Build GRCh38
rs966313873 Chr.1: 64634211-64634211 on Build GRCh38
rs199888610 Chr.1: 64634075-64634075 on Build GRCh38
rs1022855739 Chr.1: 64634091-64634091 on Build GRCh38
rs200696750 Chr.1: 64634219-64634219 on Build GRCh38
rs374615198 Chr.1: 64634243-64634243 on Build GRCh38

View Full Product Details