Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74920062-74920321 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USH1G
Entrez Gene ID
124590
Gene Name
USH1 protein network component sans
Gene Aliases
ANKS4A, SANS
UniGene
Hs.376688
Gene Chromosome Location
Chr.17: 74916083-74923263 on Build GRCh38
Transcripts
NM_001282489.2 NM_173477.4 XM_011524296.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7243366 c.591C>T p.S197S Chr.17: 74920245-74920245 on Build GRCh38
5464199 c.640A>G p.T214A Chr.17: 74920196-74920196 on Build GRCh38
1303319 c.515G>C p.R172P Chr.17: 74920321-74920321 on Build GRCh38
3521969 c.699C>T p.V233V Chr.17: 74920137-74920137 on Build GRCh38
107567 c.703G>A p.E235K Chr.17: 74920133-74920133 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs397517928 Chr.17: 74920102-74920102 on Build GRCh38
rs750109903 Chr.17: 74920237-74920237 on Build GRCh38
rs727504890 Chr.17: 74920229-74920229 on Build GRCh38
rs974350317 Chr.17: 74920256-74920256 on Build GRCh38
rs769073148 Chr.17: 74920291-74920291 on Build GRCh38
rs751372463 Chr.17: 74920247-74920247 on Build GRCh38
rs143458197 Chr.17: 74920235-74920235 on Build GRCh38
rs201123735 Chr.17: 74920273-74920273 on Build GRCh38
rs771022122 Chr.17: 74920255-74920255 on Build GRCh38
rs759953073 Chr.17: 74920108-74920108 on Build GRCh38
rs201117051 Chr.17: 74920097-74920097 on Build GRCh38
rs754390030 Chr.17: 74920209-74920209 on Build GRCh38
rs373704838 Chr.17: 74920254-74920254 on Build GRCh38
rs778772947 Chr.17: 74920124-74920124 on Build GRCh38
rs727503713 Chr.17: 74920242-74920242 on Build GRCh38
rs763002152 Chr.17: 74920099-74920099 on Build GRCh38
rs149002004 Chr.17: 74920131-74920131 on Build GRCh38
rs770350814 Chr.17: 74920200-74920201 on Build GRCh38
rs1003052325 Chr.17: 74920171-74920171 on Build GRCh38
rs202045256 Chr.17: 74920243-74920243 on Build GRCh38
rs140562408 Chr.17: 74920133-74920133 on Build GRCh38
rs759630941 Chr.17: 74920182-74920182 on Build GRCh38
rs773231689 Chr.17: 74920094-74920094 on Build GRCh38
rs756830882 Chr.17: 74920218-74920218 on Build GRCh38
rs775371036 Chr.17: 74920116-74920116 on Build GRCh38
rs779660468 Chr.17: 74920142-74920142 on Build GRCh38
rs761010255 Chr.17: 74920284-74920284 on Build GRCh38
rs774553625 Chr.17: 74920106-74920106 on Build GRCh38
rs756539436 Chr.17: 74920115-74920115 on Build GRCh38
rs779722969 Chr.17: 74920246-74920246 on Build GRCh38
rs774571563 Chr.17: 74920181-74920181 on Build GRCh38
rs777222355 Chr.17: 74920109-74920110 on Build GRCh38
rs775758795 Chr.17: 74920187-74920187 on Build GRCh38
rs146752863 Chr.17: 74920080-74920080 on Build GRCh38
rs746860881 Chr.17: 74920079-74920079 on Build GRCh38
rs3074305 Chr.17: 74920119-74920120 on Build GRCh38
rs754767231 Chr.17: 74920252-74920252 on Build GRCh38
rs544952984 Chr.17: 74920119-74920119 on Build GRCh38
rs762274233 Chr.17: 74920179-74920181 on Build GRCh38
rs774403004 Chr.17: 74920189-74920189 on Build GRCh38
rs767718565 Chr.17: 74920111-74920111 on Build GRCh38
rs200259553 Chr.17: 74920105-74920105 on Build GRCh38
rs199714230 Chr.17: 74920215-74920215 on Build GRCh38
rs907735153 Chr.17: 74920302-74920302 on Build GRCh38
rs776540260 Chr.17: 74920159-74920159 on Build GRCh38
rs886053389 Chr.17: 74920083-74920083 on Build GRCh38
rs767955984 Chr.17: 74920186-74920186 on Build GRCh38
rs558890451 Chr.17: 74920281-74920281 on Build GRCh38
rs370027457 Chr.17: 74920155-74920155 on Build GRCh38
rs200197601 Chr.17: 74920201-74920201 on Build GRCh38
rs753272581 Chr.17: 74920112-74920112 on Build GRCh38
rs201644674 Chr.17: 74920270-74920270 on Build GRCh38
rs573510359 Chr.17: 74920210-74920210 on Build GRCh38
rs530264162 Chr.17: 74920160-74920160 on Build GRCh38
rs147967199 Chr.17: 74920121-74920121 on Build GRCh38
rs201525855 Chr.17: 74920158-74920158 on Build GRCh38
rs867149581 Chr.17: 74920082-74920082 on Build GRCh38
rs770933979 Chr.17: 74920168-74920168 on Build GRCh38
rs374579438 Chr.17: 74920297-74920297 on Build GRCh38

View Full Product Details