Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 77907626-77907885 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INTS4
Entrez Gene ID
92105
Gene Name
integrator complex subunit 4
Gene Aliases
INT4, MST093
UniGene
Hs.533723
Gene Chromosome Location
Chr.11: 77874491-77994849 on Build GRCh38
Transcripts
NM_033547.3 XM_011545352.2 XM_017018560.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6134556 c.1977C>G p.F659L Chr.11: 77907756-77907756 on Build GRCh38
6462081 c.1937T>C p.L646P Chr.11: 77907796-77907796 on Build GRCh38
6134555 c.1948C>T p.Q650* Chr.11: 77907785-77907785 on Build GRCh38
245046 c.1932A>G p.Q644Q Chr.11: 77907801-77907801 on Build GRCh38
1357134 c.1942G>T p.E648* Chr.11: 77907791-77907791 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769780275 Chr.11: 77907721-77907721 on Build GRCh38
rs534107237 Chr.11: 77907740-77907740 on Build GRCh38
rs1013256436 Chr.11: 77907754-77907755 on Build GRCh38
rs1005232189 Chr.11: 77907685-77907685 on Build GRCh38
rs368646052 Chr.11: 77907841-77907841 on Build GRCh38
rs988431438 Chr.11: 77907821-77907821 on Build GRCh38
rs1014708160 Chr.11: 77907704-77907704 on Build GRCh38
rs759234854 Chr.11: 77907719-77907719 on Build GRCh38
rs866258764 Chr.11: 77907772-77907772 on Build GRCh38
rs763036024 Chr.11: 77907827-77907827 on Build GRCh38
rs143324770 Chr.11: 77907783-77907783 on Build GRCh38
rs767074781 Chr.11: 77907750-77907750 on Build GRCh38
rs367648162 Chr.11: 77907665-77907665 on Build GRCh38
rs775593919 Chr.11: 77907817-77907817 on Build GRCh38
rs761218640 Chr.11: 77907842-77907842 on Build GRCh38
rs867570154 Chr.11: 77907739-77907739 on Build GRCh38
rs781210393 Chr.11: 77907784-77907784 on Build GRCh38
rs769676358 Chr.11: 77907809-77907809 on Build GRCh38
rs147546496 Chr.11: 77907801-77907801 on Build GRCh38
rs767880416 Chr.11: 77907843-77907843 on Build GRCh38
rs748240065 Chr.11: 77907675-77907675 on Build GRCh38
rs756139774 Chr.11: 77907716-77907717 on Build GRCh38
rs956956540 Chr.11: 77907814-77907814 on Build GRCh38
rs749426544 Chr.11: 77907848-77907849 on Build GRCh38
rs747053127 Chr.11: 77907768-77907768 on Build GRCh38
rs753027868 Chr.11: 77907856-77907856 on Build GRCh38
rs147404221 Chr.11: 77907826-77907826 on Build GRCh38
rs377236226 Chr.11: 77907779-77907779 on Build GRCh38
rs773444454 Chr.11: 77907706-77907706 on Build GRCh38
rs779397386 Chr.11: 77907764-77907764 on Build GRCh38
rs772099097 Chr.11: 77907697-77907697 on Build GRCh38
rs759967316 Chr.11: 77907846-77907846 on Build GRCh38
rs758879615 Chr.11: 77907766-77907766 on Build GRCh38
rs754288588 Chr.11: 77907756-77907756 on Build GRCh38
rs755240782 Chr.11: 77907763-77907763 on Build GRCh38
rs766856862 Chr.11: 77907733-77907733 on Build GRCh38
rs199995696 Chr.11: 77907718-77907718 on Build GRCh38
rs372014766 Chr.11: 77907732-77907732 on Build GRCh38
rs368211966 Chr.11: 77907712-77907712 on Build GRCh38
rs747303681 Chr.11: 77907711-77907711 on Build GRCh38
rs557781576 Chr.11: 77907852-77907852 on Build GRCh38
rs775497743 Chr.11: 77907729-77907729 on Build GRCh38
rs759784087 Chr.11: 77907834-77907834 on Build GRCh38
rs777695636 Chr.11: 77907717-77907718 on Build GRCh38

View Full Product Details