Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 163516343-163516568 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAT2B
Entrez Gene ID
27430
Gene Name
methionine adenosyltransferase 2B
Gene Aliases
MAT-II, MATIIbeta, Nbla02999, SDR23E1, TGR
UniGene
Hs.54642
Gene Chromosome Location
Chr.5: 163503064-163519353 on Build GRCh38
Transcripts
NM_013283.4 NM_182796.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6843378 c.509G>C p.R170T Chr.5: 163516533-163516533 on Build GRCh38
5207630 c.535G>T p.V179F Chr.5: 163516526-163516526 on Build GRCh38
1696696 c.547C>T p.P183S Chr.5: 163516538-163516538 on Build GRCh38
3429225 c.577G>A p.E193K Chr.5: 163516568-163516568 on Build GRCh38
3429224 c.544G>A p.E182K Chr.5: 163516568-163516568 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs185174859 Chr.5: 163516520-163516520 on Build GRCh38
rs549855557 Chr.5: 163516509-163516509 on Build GRCh38
rs948126960 Chr.5: 163516447-163516447 on Build GRCh38
rs777456138 Chr.5: 163516433-163516435 on Build GRCh38
rs181985438 Chr.5: 163516514-163516514 on Build GRCh38
rs753502255 Chr.5: 163516480-163516480 on Build GRCh38
rs764872008 Chr.5: 163516487-163516487 on Build GRCh38
rs1016721627 Chr.5: 163516541-163516541 on Build GRCh38
rs777231225 Chr.5: 163516377-163516377 on Build GRCh38
rs760241922 Chr.5: 163516476-163516476 on Build GRCh38
rs757088282 Chr.5: 163516481-163516481 on Build GRCh38
rs760067461 Chr.5: 163516511-163516512 on Build GRCh38
rs763535591 Chr.5: 163516514-163516515 on Build GRCh38
rs763880102 Chr.5: 163516479-163516479 on Build GRCh38
rs889319468 Chr.5: 163516394-163516394 on Build GRCh38
rs984632871 Chr.5: 163516420-163516420 on Build GRCh38
rs916635450 Chr.5: 163516432-163516432 on Build GRCh38
rs548330093 Chr.5: 163516463-163516463 on Build GRCh38
rs771333662 Chr.5: 163516523-163516523 on Build GRCh38
rs931294113 Chr.5: 163516411-163516411 on Build GRCh38
rs371182015 Chr.5: 163516525-163516525 on Build GRCh38
rs758322889 Chr.5: 163516506-163516506 on Build GRCh38
rs767500924 Chr.5: 163516381-163516381 on Build GRCh38
rs763480646 Chr.5: 163516542-163516542 on Build GRCh38
rs750226361 Chr.5: 163516497-163516497 on Build GRCh38
rs368385582 Chr.5: 163516515-163516515 on Build GRCh38
rs749416883 Chr.5: 163516516-163516516 on Build GRCh38
rs1046572948 Chr.5: 163516464-163516464 on Build GRCh38

View Full Product Details