Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154273637-154273862 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAX1
Entrez Gene ID
10456
Gene Name
HCLS1 associated protein X-1
Gene Aliases
HCLSBP1, HS1BP1, SCN3
UniGene
Hs.199625
Gene Chromosome Location
Chr.1: 154272563-154275875 on Build GRCh38
Transcripts
NM_001018837.1 NM_006118.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4022774 c.359G>A p.R120Q Chr.1: 154273816-154273816 on Build GRCh38
675882 c.385A>G p.M129V Chr.1: 154273842-154273842 on Build GRCh38
6677788 c.376C>T p.R126W Chr.1: 154273833-154273833 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs762197407 Chr.1: 154273814-154273814 on Build GRCh38
rs1014791330 Chr.1: 154273757-154273757 on Build GRCh38
rs780048656 Chr.1: 154273727-154273727 on Build GRCh38
rs904794613 Chr.1: 154273665-154273665 on Build GRCh38
rs367673863 Chr.1: 154273796-154273796 on Build GRCh38
rs1043353596 Chr.1: 154273662-154273662 on Build GRCh38
rs371368602 Chr.1: 154273785-154273785 on Build GRCh38
rs886045320 Chr.1: 154273815-154273815 on Build GRCh38
rs910542080 Chr.1: 154273708-154273708 on Build GRCh38
rs751629934 Chr.1: 154273736-154273736 on Build GRCh38
rs754267551 Chr.1: 154273679-154273679 on Build GRCh38
rs943409929 Chr.1: 154273724-154273724 on Build GRCh38
rs188953441 Chr.1: 154273747-154273747 on Build GRCh38
rs758528569 Chr.1: 154273722-154273722 on Build GRCh38
rs765450364 Chr.1: 154273702-154273702 on Build GRCh38
rs777055928 Chr.1: 154273807-154273807 on Build GRCh38
rs765538745 Chr.1: 154273833-154273833 on Build GRCh38
rs147593581 Chr.1: 154273741-154273741 on Build GRCh38
rs764630660 Chr.1: 154273669-154273669 on Build GRCh38
rs147036748 Chr.1: 154273834-154273834 on Build GRCh38
rs1053707132 Chr.1: 154273815-154273815 on Build GRCh38
rs371504152 Chr.1: 154273772-154273772 on Build GRCh38
rs368305272 Chr.1: 154273770-154273770 on Build GRCh38
rs375533183 Chr.1: 154273769-154273769 on Build GRCh38
rs758969838 Chr.1: 154273661-154273661 on Build GRCh38
rs762152245 Chr.1: 154273694-154273694 on Build GRCh38
rs374758765 Chr.1: 154273794-154273794 on Build GRCh38
rs778735894 Chr.1: 154273755-154273755 on Build GRCh38
rs933251488 Chr.1: 154273667-154273667 on Build GRCh38
rs17851424 Chr.1: 154273793-154273793 on Build GRCh38
rs756075374 Chr.1: 154273739-154273739 on Build GRCh38
rs750604799 Chr.1: 154273712-154273712 on Build GRCh38

View Full Product Details