Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 69209568-69209823 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXD2
Entrez Gene ID
55218
Gene Name
exonuclease 3'-5' domain containing 2
Gene Aliases
C14orf114, EXDL2
UniGene
Hs.533878
Gene Chromosome Location
Chr.14: 69191477-69244020 on Build GRCh38
Transcripts
NM_001193360.1 NM_001193361.1 NM_001193362.1 NM_001193363.1 NM_018199.3 NR_034164.1 NR_034165.1 XM_011536908.2 XM_017021421.1

Gene Symbol
LOC105370550
Entrez Gene ID
105370550
Gene Name
uncharacterized LOC105370550
Gene Aliases
-
UniGene
Hs.667208
Gene Chromosome Location
Chr.14: 69183018-69260567 on Build GRCh38
Transcripts
NM_001193360.1 NM_001193361.1 NM_001193362.1 NM_001193363.1 NM_018199.3 NR_034164.1 NR_034165.1 XM_011536908.2 XM_017021421.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6194857 c.318A>C p.G106G Chr.14: 69209788-69209788 on Build GRCh38
239753 c.125A>G p.Q42R Chr.14: 69209595-69209595 on Build GRCh38
6333400 c.261G>T p.E87D Chr.14: 69209731-69209731 on Build GRCh38
5522611 c.301G>A p.D101N Chr.14: 69209771-69209771 on Build GRCh38
5740148 c.113C>A p.P38H Chr.14: 69209583-69209583 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749547416 Chr.14: 69209747-69209747 on Build GRCh38
rs751529492 Chr.14: 69209656-69209656 on Build GRCh38
rs902507379 Chr.14: 69209790-69209790 on Build GRCh38
rs556197203 Chr.14: 69209677-69209677 on Build GRCh38
rs1014072150 Chr.14: 69209721-69209721 on Build GRCh38
rs756354734 Chr.14: 69209743-69209743 on Build GRCh38
rs763017009 Chr.14: 69209771-69209771 on Build GRCh38
rs763872672 Chr.14: 69209640-69209640 on Build GRCh38
rs757322538 Chr.14: 69209659-69209659 on Build GRCh38
rs779199657 Chr.14: 69209777-69209777 on Build GRCh38
rs1016650883 Chr.14: 69209705-69209705 on Build GRCh38
rs1004874222 Chr.14: 69209676-69209676 on Build GRCh38
rs766419235 Chr.14: 69209658-69209658 on Build GRCh38
rs1028277326 Chr.14: 69209762-69209762 on Build GRCh38
rs762841088 Chr.14: 69209636-69209636 on Build GRCh38
rs75083088 Chr.14: 69209744-69209744 on Build GRCh38
rs752389207 Chr.14: 69209598-69209598 on Build GRCh38
rs963430013 Chr.14: 69209713-69209713 on Build GRCh38
rs750579457 Chr.14: 69209688-69209688 on Build GRCh38
rs769075965 Chr.14: 69209749-69209749 on Build GRCh38
rs895674480 Chr.14: 69209715-69209715 on Build GRCh38
rs537950944 Chr.14: 69209669-69209669 on Build GRCh38
rs764922126 Chr.14: 69209590-69209590 on Build GRCh38

View Full Product Details