Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 102934520-102934775 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL13
Entrez Gene ID
222235
Gene Name
F-box and leucine rich repeat protein 13
Gene Aliases
DRC6, Fbl13
UniGene
Hs.660029
Gene Chromosome Location
Chr.7: 102699226-103074841 on Build GRCh38
Transcripts
NM_001111038.1 NM_001287150.1 NM_145032.3 NR_105043.1 XM_005250205.3 XM_005250207.3 XM_005250208.3 XM_005250209.2 XM_006715898.2 XM_011515928.2 XM_011515929.2 XM_011515930.2 XM_011515932.2 XM_017011850.1 XM_017011851.1 XM_017011852.1 XM_017011853.1 NM_001031692.2 NM_005824.2 XM_005250108.1

Gene Symbol
LRRC17
Entrez Gene ID
10234
Gene Name
leucine rich repeat containing 17
Gene Aliases
P37NB
UniGene
Hs.567412
Gene Chromosome Location
Chr.7: 102912897-102945111 on Build GRCh38
Transcripts
NM_001111038.1 NM_001287150.1 NM_145032.3 NR_105043.1 XM_005250205.3 XM_005250207.3 XM_005250208.3 XM_005250209.2 XM_006715898.2 XM_011515928.2 XM_011515929.2 XM_011515930.2 XM_011515932.2 XM_017011850.1 XM_017011851.1 XM_017011852.1 XM_017011853.1 NM_001031692.2 NM_005824.2 XM_005250108.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7308343 c.646G>A p.E216K Chr.7: 102934559-102934559 on Build GRCh38
3631468 c.672C>T p.P224P Chr.7: 102934585-102934585 on Build GRCh38
1083526 c.731A>G p.N244S Chr.7: 102934644-102934644 on Build GRCh38
6426372 c.642A>T p.E214D Chr.7: 102934555-102934555 on Build GRCh38
6193561 c.702G>T p.K234N Chr.7: 102934615-102934615 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200274905 Chr.7: 102934579-102934579 on Build GRCh38
rs768188544 Chr.7: 102934698-102934701 on Build GRCh38
rs755652219 Chr.7: 102934578-102934578 on Build GRCh38
rs753292751 Chr.7: 102934674-102934674 on Build GRCh38
rs375932026 Chr.7: 102934551-102934551 on Build GRCh38
rs148357310 Chr.7: 102934606-102934606 on Build GRCh38
rs748325991 Chr.7: 102934724-102934724 on Build GRCh38
rs80100722 Chr.7: 102934559-102934559 on Build GRCh38
rs919990646 Chr.7: 102934596-102934596 on Build GRCh38
rs773193596 Chr.7: 102934615-102934615 on Build GRCh38
rs146334968 Chr.7: 102934627-102934627 on Build GRCh38
rs762878029 Chr.7: 102934619-102934619 on Build GRCh38
rs113237255 Chr.7: 102934560-102934560 on Build GRCh38
rs1047256967 Chr.7: 102934613-102934613 on Build GRCh38
rs141262991 Chr.7: 102934607-102934607 on Build GRCh38
rs77576273 Chr.7: 102934555-102934555 on Build GRCh38
rs755854852 Chr.7: 102934658-102934658 on Build GRCh38
rs780414807 Chr.7: 102934692-102934692 on Build GRCh38
rs557836390 Chr.7: 102934717-102934717 on Build GRCh38
rs574386315 Chr.7: 102934655-102934655 on Build GRCh38
rs377676475 Chr.7: 102934622-102934622 on Build GRCh38
rs752374293 Chr.7: 102934648-102934648 on Build GRCh38
rs3832497 Chr.7: 102934553-102934555 on Build GRCh38
rs751312990 Chr.7: 102934546-102934546 on Build GRCh38
rs36040681 Chr.7: 102934571-102934571 on Build GRCh38
rs141994496 Chr.7: 102934609-102934609 on Build GRCh38
rs372298144 Chr.7: 102934550-102934550 on Build GRCh38
rs746338531 Chr.7: 102934589-102934589 on Build GRCh38
rs758886612 Chr.7: 102934683-102934683 on Build GRCh38
rs150294157 Chr.7: 102934642-102934642 on Build GRCh38
rs757868779 Chr.7: 102934601-102934601 on Build GRCh38
rs531650613 Chr.7: 102934559-102934561 on Build GRCh38
rs772488729 Chr.7: 102934597-102934597 on Build GRCh38
rs780398889 Chr.7: 102934598-102934598 on Build GRCh38
rs762509863 Chr.7: 102934551-102934553 on Build GRCh38
rs752043474 Chr.7: 102934594-102934597 on Build GRCh38
rs190412119 Chr.7: 102934645-102934645 on Build GRCh38
rs747585651 Chr.7: 102934705-102934705 on Build GRCh38
rs769909320 Chr.7: 102934726-102934726 on Build GRCh38
rs769069587 Chr.7: 102934602-102934602 on Build GRCh38
rs750768621 Chr.7: 102934704-102934709 on Build GRCh38
rs536003329 Chr.7: 102934601-102934601 on Build GRCh38
rs777298387 Chr.7: 102934663-102934663 on Build GRCh38
rs73408251 Chr.7: 102934753-102934753 on Build GRCh38

View Full Product Details