Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 101185430-101185685 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLPPR1
Entrez Gene ID
54886
Gene Name
phospholipid phosphatase related 1
Gene Aliases
LPPR1, PRG-3
UniGene
Hs.382683
Gene Chromosome Location
Chr.9: 101028749-101325135 on Build GRCh38
Transcripts
NM_017753.2 NM_207299.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1458944 c.55T>C p.F19L Chr.9: 101185549-101185549 on Build GRCh38
7375382 c.34T>G p.S12A Chr.9: 101185528-101185528 on Build GRCh38
107371 c.42C>T p.I14I Chr.9: 101185536-101185536 on Build GRCh38
4494809 c.43C>T p.P15S Chr.9: 101185537-101185537 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1054472662 Chr.9: 101185615-101185615 on Build GRCh38
rs146000696 Chr.9: 101185548-101185548 on Build GRCh38
rs772068064 Chr.9: 101185598-101185598 on Build GRCh38
rs1033618675 Chr.9: 101185576-101185576 on Build GRCh38
rs533287497 Chr.9: 101185488-101185488 on Build GRCh38
rs776868648 Chr.9: 101185600-101185600 on Build GRCh38
rs766404145 Chr.9: 101185481-101185481 on Build GRCh38
rs377630329 Chr.9: 101185486-101185486 on Build GRCh38
rs537461048 Chr.9: 101185640-101185640 on Build GRCh38
rs377112102 Chr.9: 101185492-101185492 on Build GRCh38
rs370807466 Chr.9: 101185487-101185487 on Build GRCh38
rs760462653 Chr.9: 101185617-101185617 on Build GRCh38
rs142421575 Chr.9: 101185539-101185539 on Build GRCh38
rs75832966 Chr.9: 101185502-101185502 on Build GRCh38
rs747826350 Chr.9: 101185459-101185459 on Build GRCh38
rs774148244 Chr.9: 101185588-101185588 on Build GRCh38
rs928324444 Chr.9: 101185510-101185510 on Build GRCh38
rs200860367 Chr.9: 101185520-101185520 on Build GRCh38
rs760678230 Chr.9: 101185603-101185604 on Build GRCh38
rs759578933 Chr.9: 101185589-101185589 on Build GRCh38
rs975694691 Chr.9: 101185636-101185636 on Build GRCh38
rs769492694 Chr.9: 101185545-101185545 on Build GRCh38
rs751702937 Chr.9: 101185495-101185495 on Build GRCh38
rs879080960 Chr.9: 101185620-101185620 on Build GRCh38
rs568909287 Chr.9: 101185635-101185635 on Build GRCh38
rs34705940 Chr.9: 101185603-101185603 on Build GRCh38
rs762641094 Chr.9: 101185475-101185475 on Build GRCh38
rs770611688 Chr.9: 101185561-101185561 on Build GRCh38
rs765501670 Chr.9: 101185509-101185509 on Build GRCh38
rs747599995 Chr.9: 101185541-101185541 on Build GRCh38
rs41296085 Chr.9: 101185528-101185528 on Build GRCh38
rs769369805 Chr.9: 101185466-101185466 on Build GRCh38
rs773071707 Chr.9: 101185467-101185467 on Build GRCh38
rs749146107 Chr.9: 101185560-101185560 on Build GRCh38
rs566713852 Chr.9: 101185518-101185518 on Build GRCh38
rs773434155 Chr.9: 101185596-101185597 on Build GRCh38
rs1023937851 Chr.9: 101185534-101185534 on Build GRCh38
rs143808898 Chr.9: 101185536-101185536 on Build GRCh38
rs369750856 Chr.9: 101185606-101185606 on Build GRCh38

View Full Product Details